    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Tensin 4 Antibodies
          Invitrogen
          Tensin 4 Recombinant Rabbit Monoclonal Antibody (K01_2J41)
          View all (14) Tensin 4 antibodies
          Questions & Answers
          Cite Tensin 4 Recombinant Rabbit Monoclonal Antibody (K01_2J41)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          Tensin 4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Tensin 4 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Tensin 4 Antibody (MA563880) in WB
          Western blot analysis of Tensin 4 in various lysates. Samples were incubated with Tensin 4 recombinant monoclonal (Product # MA563880). Samples: Raw264.7, C6 lysates. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Tensin 4 Antibody in Western Blot (WB)
          Tensin 4 Antibody in Western Blot (WB)
          Product Details
          MA563880
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_2J41
          Immunogen
          A synthetic peptide of human TNS4.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          50mM tris glycine, pH 7.4, with 40% glycerol, 0.05% BSA, 150mM NaCl
          Contains
          0.01% sodium azide
          Storage conditions
          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3704928
          Target Information
          TNS4 links the focal adhesion and cell adhesion signaling to the actin cytoskeleton. It may be involved in cell survival and migration. The protein may be involved in cell migration, cartilage development and in linking signal transduction pathways to the cytoskeleton and promote apoptosis, via its cleavage by caspase-3.
          Bioinformatics
          Protein Aliases: C terminal tensin like; c-ten; C-terminal tensin-like protein; cten; Tensin-4; unnamed protein product
          View more View less
          Gene Aliases: 9930017A07Rik; AA589547; AU016405; CTEN; PP14434; RGD1310402; TNS4
          View more View less
          UniProt ID: (Human) Q8IZW8, (Rat) Q4V8I3, (Mouse) Q8BZ33
          View more View less
          Entrez Gene ID: (Human) 84951, (Rat) 303517, (Mouse) 217169
          View more View less
          Function(s)
          actin binding protein binding molecular_function non-motor actin binding protein
          Process(es)
          protein localization apoptotic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline