    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HJURP Antibodies
          Invitrogen
          HJURP Recombinant Rabbit Monoclonal Antibody (K01_2M72)
          View all (14) HJURP antibodies
          Questions & Answers
          Cite HJURP Recombinant Rabbit Monoclonal Antibody (K01_2M72)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          HJURP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          HJURP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          HJURP Antibody (MA564086) in IHC (P)
          Immunohistochemistry (Paraffin) analysis of HJURP in paraffin-embedded Human lung cancer tissue. Samples were incubated with HJURP recombinant monoclonal (Product # MA564086). High-pressure and temperature Sodium Citrate pH 6.0 was used for antigen retrieval. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HJURP Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HJURP Antibody in Western Blot (WB)
          Product Details
          MA564086
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:100
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_2M72
          Immunogen
          Recombinant protein of human HJURP.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          50mM tris glycine, pH 7.4, with 40% glycerol, 150mM NaCl, 0.05% BSA
          Contains
          0.01% sodium azide
          Storage conditions
          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3705134
          Target Information
          Holliday junction recognition protein (HJURP) is a centromeric protein that plays a central role in the incorporation and maintenance of histone H3-like variant CENPA at centromeres. It acts as a specific chaperone for CENPA and is required for the incorporation of newly synthesized CENPA molecules into nucleosomes at replicated centromeres. HJURP prevents CENPA-H4 tetramerization and prevents premature DNA binding by the CENPA-H4 tetramer. It directly binds to Holliday junctions.
          Bioinformatics
          Protein Aliases: 14-3-3-associated AKT substrate; fetal liver expressing gene 1; Fetal liver-expressing gene 1 protein; hFLEG1; Holliday junction recognition protein; unnamed protein product; Up-regulated in lung cancer 9
          View more View less
          Gene Aliases: FAKTS; FLEG1; hFLEG1; HJURP; URLC9
          View more View less
          UniProt ID: (Human) Q8NCD3
          View more View less
          Entrez Gene ID: (Human) 55355
          View more View less
          Function(s)
          DNA binding protein binding histone binding identical protein binding
          Process(es)
          chromosome segregation CENP-A containing nucleosome assembly regulation of protein complex assembly regulation of DNA binding
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline