    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Glyphosate Antibodies
          Invitrogen
          Glyphosate Recombinant Mouse Monoclonal Antibody (3F10)
          View all (2) Glyphosate antibodies
          Questions & Answers
          Cite Glyphosate Recombinant Mouse Monoclonal Antibody (3F10)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Glyphosate Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          Glyphosate Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Glyphosate Antibody (MA566470) in SDS-PAGE
          SDS-PAGE characterization of Glyphosate under reducing conditions. Lane M: Protein marker (kDa); Lane GLP: Purified Glyphosate Recombinant Monoclonal (Product # MA566470). Glyphosate showing heavy chain (~50 kDa) and light chain (~25 kDa). Gels were stained with Coomassie Brilliant Blue R-595. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Glyphosate Antibody in SDS-PAGE (SDS-PAGE)
          Product Details
          MA566470
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:5,000
          -
          Product Specifications
          Species Reactivity
          Chemical
          Host/Isotype
          Mouse / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          3F10
          Immunogen
          Conjugate of GLP hapten and KLH.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          7 mg/mL
          Amount
          200 µg
          Purification
          Protein G
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3715617
          Target Information
          Glyphosate is a broad-spectrum systemic herbicide and crop desiccant, meaning it will kill most plants. It inhibits plant enzyme 5-enolpyruvylshikimate-3-phosphate synthase in the shikimic acid pathway. The shikimic acid pathway is necessary for plants and some microorganisms.
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline