    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Fluazinam Antibodies
          Invitrogen
          Fluazinam Recombinant Mouse Monoclonal Antibody (2G9)
          Questions & Answers
          Cite Fluazinam Recombinant Mouse Monoclonal Antibody (2G9)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Fluazinam Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          Fluazinam Antibody in SDS-PAGE (SDS-PAGE)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Fluazinam Antibody (MA566494) in SDS-PAGE
          SDS-PAGE characterization of Fluazinam under reducing conditions. Lane M: Protein marker (kDa); Lane FZN: Purified Fluazinam Recombinant Monoclonal (Product # MA566494). Fluazinam showing heavy chain (~50 kDa) and light chain (~25 kDa). Gels were stained with Coomassie Brilliant Blue R-514. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Fluazinam Antibody in SDS-PAGE (SDS-PAGE)
          Product Details
          MA566494
          Applications
          Tested Dilution
          Publications
          ELISA (ELISA)
          1:5,000
          -
          Product Specifications
          Species Reactivity
          Chemical
          Host/Isotype
          Mouse / IgG
          Expression System
          CHO cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          2G9
          Immunogen
          Conjugate of FZN hapten and KLH.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          3.7 mg/mL
          Amount
          200 µg
          Purification
          Protein G
          Storage buffer
          PBS
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_3715641
          Target Information
          Fluazinam is a broad-spectrum fungicide belonging to the pyrimidine class, widely used in agriculture to control various fungal diseases such as late blight, gray mold, and clubroot across a range of crops including potatoes, vegetables, and ornamental plants. Structurally, fluazinam features a pyrimidine ring linked to a nitro group, which facilitates its role as an inhibitor of fungal respiration by uncoupling oxidative phosphorylation in mitochondria, thereby disrupting energy production in target pathogens. Fluazinam is effective as both a protectant and curative treatment, with applications typically via foliar spray or soil treatment to ensure thorough coverage and absorption. Its persistent efficacy and low risk of resistance development make it valuable in integrated pest management programs, although environmental considerations regarding non-target aquatic organisms necessitate cautious application near water bodies. Research continues to explore its environmental behavior and develop guidelines for safe usage to optimize its impact while minimizing ecological disruption.
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0