    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • BAZ1B Antibodies
          Invitrogen
          BAZ1B Recombinant Rabbit Monoclonal Antibody (K01_3P13)
          View all (18) BAZ1B antibodies
          Questions & Answers
          Cite BAZ1B Recombinant Rabbit Monoclonal Antibody (K01_3P13)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          BAZ1B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BAZ1B Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          BAZ1B Antibody (MA566753) in WB
          Western Blot analysis of BAZ1B in SH-SY5Y lysates using BAZ1B Recombinant Monoclonal Antibody (Product # MA566753). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          BAZ1B Antibody in Western Blot (WB)
          Product Details
          MA566753
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_3P13
          Immunogen
          A synthesized peptide derived from human WSTF.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA, 50% glycerol, 150mM NaCl
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          BAZ1B is a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.This gene encodes a member of the bromodomain protein family. The bromodomain is a structural motif characteristic of proteins involved in chromatin-dependent regulation of transcription. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
          Bioinformatics
          Protein Aliases: Bromodomain adjacent to zinc finger domain protein 1B; bromodomain adjacent to zinc finger domain, 1B; Williams-Beuren syndrome chromosome region 9 homolog; hWALp2; presumptive transcriptional regulator; transcription factor WSTF; Tyrosine-protein kinase BAZ1B; Williams syndrome transcription factor; Williams syndrome transcription factor homolog; Williams-Beuren syndrome chromosomal region 10 protein; Williams-Beuren syndrome chromosomal region 9 protein; Williams-Beuren syndrome chromosomal region 9 protein homolog; Williams-Beuren syndrome chromosome region 9 homolog
          View more View less
          Gene Aliases: BAZ1B; C87820; WBSC10; WBSCR10; WBSCR9; WSTF
          View more View less
          UniProt ID: (Human) Q9UIG0, (Mouse) Q9Z277
          View more View less
          Entrez Gene ID: (Human) 9031, (Rat) 368002, (Mouse) 22385
          View more View less
          Function(s)
          nucleotide binding protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity protein binding ATP binding zinc ion binding kinase activity transferase activity histone kinase activity histone binding metal ion binding histone H2AXY142 kinase activity molecular_function chromatin binding lysine-acetylated histone binding vitamin D receptor activator activity
          Process(es)
          heart morphogenesis double-strand break repair chromatin assembly or disassembly chromatin remodeling regulation of transcription, DNA-templated cellular response to DNA damage stimulus biological_process histone phosphorylation peptidyl-tyrosine phosphorylation chromatin-mediated maintenance of transcription positive regulation of receptor activity chromatin organization regulation of transcription from RNA polymerase II promoter post-translational protein modification positive regulation of transcription from RNA polymerase I promoter positive regulation of transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase III promoter negative regulation of mitotic chromosome condensation transcription, DNA-templated phosphorylation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline