    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Torsin 1A Antibodies
          Invitrogen
          Torsin 1A Recombinant Rabbit Monoclonal Antibody (K01_3R76)
          View all (10) Torsin 1A antibodies
          Questions & Answers
          Cite Torsin 1A Recombinant Rabbit Monoclonal Antibody (K01_3R76)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Torsin 1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Torsin 1A Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Torsin 1A Antibody (MA566900) in WB
          Western Blot analysis of Torsin 1A in 293T lysates using Torsin 1A Recombinant Monoclonal Antibody (Product # MA566900). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Torsin 1A Antibody in Western Blot (WB)
          Product Details
          MA566900
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_3R76
          Immunogen
          A synthesized peptide derived from human Torsin A.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 50% glycerol, 150mM NaCl, 0.05% BSA
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          The protein encoded by this gene is a member of the AAA family of adenosine triphosphatases (ATPases), is related to the Clp protease/heat shock family and is expressed prominently in the substantia nigra pars compacta. Mutations in this gene result in the autosomal dominant disorder, torsion dystonia 1.
          Bioinformatics
          Protein Aliases: dystonia 1; Dystonia 1 protein; Dystonia 1 torsion; dystonia 1, torsion (autosomal dominant; torsin A); Torsin 1A; torsin A; torsin ATPase 1; Torsin ATPase-1A; Torsin family 1 member A; Torsin-1A; unnamed protein product
          View more View less
          Gene Aliases: AMC5; DQ2; DYT1; TA; TOR1A; TORA
          View more View less
          UniProt ID: (Human) O14656
          View more View less
          Entrez Gene ID: (Human) 1861
          View more View less
          Function(s)
          nucleotide binding protein binding ATP binding cytoskeletal protein binding hydrolase activity ATPase activity kinesin binding identical protein binding unfolded protein binding misfolded protein binding ATP-dependent protein folding chaperone
          Process(es)
          protein deneddylation protein folding response to oxidative stress nuclear envelope organization cell adhesion neuron projection development protein localization to nucleus ERAD pathway wound healing, spreading of cells intermediate filament cytoskeleton organization synaptic vesicle transport synaptic vesicle membrane organization regulation of dopamine uptake involved in synaptic transmission nuclear membrane organization positive regulation of synaptic vesicle endocytosis regulation of protein localization to cell surface
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline