    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Reelin Antibodies
          Invitrogen
          Reelin Recombinant Rabbit Monoclonal Antibody (K01_3U15)
          View all (15) Reelin antibodies
          Questions & Answers
          Cite Reelin Recombinant Rabbit Monoclonal Antibody (K01_3U15)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Reelin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Reelin Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Reelin Antibody (MA567000) in WB
          Western Blot analysis of Reelin in HepG2 lysates using Reelin Recombinant Monoclonal Antibody (Product # MA567000). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Reelin Antibody in Western Blot (WB)
          Product Details
          MA567000
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_3U15
          Immunogen
          A synthesized peptide derived from human Reelin.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA, 50% glycerol, 150mM NaCl
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Reelin in an extracellular matrix serine protease that plays a role in layering of neurons in the cerebral cortex and cerebellum. Regulates microtubule function in neurons and neuronal migration. Affects migration of sympathetic preganglionic neurons in the spinal cord, where it seems to act as a barrier to neuronal migration. Enzymatic activity is important for the modulation of cell adhesion. Binding to the extracellular domains of lipoprotein receptors VLDLR and LRP8/APOER2 induces tyrosine phosphorylation of DAB1 and modulation of TAU phosphorylation.
          Bioinformatics
          Protein Aliases: Reelin; unnamed protein product
          View more View less
          Gene Aliases: ETL7; LIS2; PRO1598; Reelen; reeler; RELN; RL
          View more View less
          UniProt ID: (Human) P78509, (Rat) P58751
          View more View less
          Entrez Gene ID: (Human) 5649, (Rat) 24718
          View more View less
          Function(s)
          peptidase activity serine-type peptidase activity hydrolase activity metal ion binding receptor agonist activity lipoprotein particle receptor binding very-low-density lipoprotein particle receptor binding molecular_function protein serine/threonine/tyrosine kinase activity extracellular matrix glycoprotein extracellular matrix protein
          Process(es)
          cell morphogenesis neuron migration proteolysis cell adhesion axon guidance central nervous system development brain development learning long-term memory locomotory behavior associative learning glial cell differentiation regulation of gene expression positive regulation of neuron projection development dendrite development cell migration spinal cord patterning ventral spinal cord development hippocampus development cerebral cortex tangential migration layer formation in cerebral cortex cerebral cortex development forebrain development positive regulation of TOR signaling protein localization to synapse reelin-mediated signaling pathway proteasome-mediated ubiquitin-dependent protein catabolic process regulation of neuron differentiation response to pain regulation of behavior modulation of synaptic transmission positive regulation of small GTPase mediated signal transduction positive regulation of protein kinase B signaling positive regulation of synaptic transmission, glutamatergic regulation of synaptic activity long-term synaptic potentiation positive regulation of dendritic spine morphogenesis regulation of synapse maturation positive regulation of synapse maturation postsynaptic density assembly NMDA glutamate receptor clustering postsynaptic density protein 95 clustering receptor localization to synapse motor neuron migration lateral motor column neuron migration positive regulation of long-term synaptic potentiation positive regulation of lateral motor column neuron migration interneuron migration positive regulation of excitatory postsynaptic potential regulation of neuron migration cell morphogenesis involved in differentiation midgut development memory response to mechanical stimulus response to toxic substance response to hormone response to lithium ion positive regulation of phosphatidylinositol 3-kinase signaling response to organic cyclic compound response to insecticide peptidyl-tyrosine phosphorylation dentate gyrus development orbitofrontal cortex development hypothalamus development response to progesterone positive regulation of CREB transcription factor activity social behavior thyroid hormone metabolic process response to drug response to amino acid response to ethanol positive regulation of protein kinase activity response to axon injury positive regulation of peptidyl-tyrosine phosphorylation response to corticosterone response to electrical stimulus positive regulation of protein tyrosine kinase activity response to fatty acid cellular response to dexamethasone stimulus regulation of N-methyl-D-aspartate selective glutamate receptor activity positive regulation of alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionate selective glutamate receptor activity
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline