    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TAOK1/TAOK2/TAOK3 Antibodies
          Invitrogen
          Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Recombinant Rabbit Monoclonal Antibody (K01_4Q66)
          View all (3) TAOK1/TAOK2/TAOK3 antibodies
          Questions & Answers
          Cite Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Recombinant Rabbit Monoclonal Antibody (K01_4Q66)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Antibody (MA567642) in WB
          Western Blot analysis of Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) in Lane 1: 293 lysates; Lane 2: 293 cell treated with Lambda Phosphatase lysates using Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Recombinant Monoclonal Antibody (Product # MA567642). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Antibody in Western Blot (WB)
          Phospho-TAOK1/TAOK2/TAOK3 (Ser181, Ser177) Recombinant Rabbit Monoclonal Antibody (K01_4Q66)
          Product Details
          MA567642
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_4Q66
          Immunogen
          A synthesized peptide derived from human Phospho-TAOK1/2/3 (S181 + S181 + S177).
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          PBS, pH 7.4, with 0.05% BSA, 150mM NaCl, 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          TAOK kinases, including TAOK1, TAOK2, and TAOK3, are serine-threonine kinases that belong to the sterile 20 (STE20) family and play significant roles in various cellular processes, including proliferation, apoptosis, and tumor progression. TAOK1 is implicated in neurodevelopmental disorders and has an evolving phenotype and genotype spectrum, which includes associations with macrocephaly and autism spectrum disorders. The TAOK2 gene, located on chromosome 16, is a crucial kinase participating in the regulation of protein translation through the phosphorylation of eukaryotic elongation factor 2 (eEF2) and is also involved in neurodevelopmental pathways. Moreover, TAOK2 acts as a potential tumor suppressor, with its downregulation linked to cancer progression in numerous types of cancer. TAOK3, another member of this kinase family, is expressed ubiquitously and has been linked to the regulation of age-associated inflammation, acting through pathways that limit pro-inflammatory monocyte accumulation. Additionally, TAOK3 is involved in osteoblast differentiation and modulates T cell receptor signaling by influencing components like SHP-1 and LCK. All three kinases are crucial for maintaining cellular homeostasis and their aberrations can lead to pathological conditions, particularly related to neurodevelopment and cancer.
          Bioinformatics
          Protein Aliases: Axotomy-related gene 357 protein; CTCL-associated antigen HD-CL-09; Cutaneous T-cell lymphoma-associated antigen HD-CL-09; Dendritic cell-derived protein kinase; hKFC-A; hKFC-B; hKFC-C; JNK/SAPK-inhibitory kinase; Jun kinase-inhibitory kinase; KDS; Kinase from chicken homolog A; Kinase from chicken homolog B; Kinase from chicken homolog C; MARK Kinase; MARKK; microtubule affinity regulating kinase kinase; prostate derived STE20-like kinase PSK; prostate-derived STE20-like kinase 1; prostate-derived STE20-like kinase 2; Prostate-derived sterile 20-like kinase 1; Prostate-derived sterile 20-like kinase 2; protein kinase homolog; PSK-1; PSK-2; serine-threonine kinase; serine/threonine protein kinase TAO1; serine/threonine protein kinase TAO1 homolog; serine/threonine protein kinase TAO2; Serine/threonine-protein kinase TAO1; Serine/threonine-protein kinase TAO2; Serine/threonine-protein kinase TAO3; STE20-like kinase; STE20-like kinase PSK2; TAO kinase 2TAO kinase 2; TAOK1; Thousand and one amino acid protein 1; Thousand and one amino acid protein 2; Thousand and one amino acid protein 3; Thousand and one amino acid protein kinase 1; Thousand and one amino acid protein kinase 2; unnamed protein product
          View more View less
          Gene Aliases: 1110033K02Rik; 2810468K05Rik; 2900006A08Rik; A130052D22; A430105I05Rik; Arg357; AU020252; B230344N16; D130018F14Rik; DDIB; DPK; hKFC-A; hKFC-B; hTAOK1; JIK; KDS; KFC-B; KIAA0881; KIAA1361; MAP3K16; MAP3K17; MAP3K18; MARKK; mKIAA0881; mKIAA1361; PSK; PSK-2; PSK1; PSK1-BETA; PSK2; TAO1; TAO2; Tao2beta; TAOK1; TAOK2; TAOK3; UNQ2971/PRO7431
          View more View less
          UniProt ID: (Human) Q7L7X3, (Human) Q9UL54, (Human) Q9H2K8, (Mouse) Q5F2E8, (Rat) O88664, (Mouse) Q6ZQ29, (Rat) Q9JLS3, (Mouse) Q8BYC6, (Rat) Q53UA7
          View more View less
          Entrez Gene ID: (Human) 57551, (Human) 9344, (Human) 51347, (Mouse) 216965, (Rat) 286993, (Mouse) 381921, (Rat) 64666, (Mouse) 330177, (Rat) 304530
          View more View less
          Function(s)
          nucleotide binding protein kinase activity protein serine/threonine kinase activity protein binding ATP binding kinase activity transferase activity alpha-tubulin binding protein serine/threonine kinase activator activity tau protein binding beta-tubulin binding tau-protein kinase activity protein serine kinase activity MAP kinase kinase kinase activity mitogen-activated protein kinase kinase binding neuropilin binding protein kinase inhibitor activity transferase activity, transferring phosphorus-containing groups protein kinase binding protein kinase activator activity non-receptor serine/threonine protein kinase
          Process(es)
          microtubule cytoskeleton organization DNA repair apoptotic process cellular response to DNA damage stimulus negative regulation of microtubule depolymerization mitotic G2 DNA damage checkpoint central nervous system neuron development positive regulation of stress-activated MAPK cascade regulation of actin cytoskeleton organization positive regulation of JNK cascade regulation of cytoskeleton organization neuron cellular homeostasis regulation of microtubule cytoskeleton organization execution phase of apoptosis regulation of cell growth protein targeting to membrane axonogenesis regulation of cell shape cell migration actin cytoskeleton organization positive regulation of MAPK cascade focal adhesion assembly cell morphogenesis involved in neuron differentiation neuron projection morphogenesis stress-activated MAPK cascade basal dendrite morphogenesis basal dendrite arborization MAPK cascade protein phosphorylation regulation of MAPK cascade positive regulation of JUN kinase activity negative regulation of JNK cascade activation of MAPKK activity regulation of mitotic cell cycle phosphorylation signal transduction by protein phosphorylation stress-activated protein kinase signaling cascade G2 DNA damage checkpoint regulation of apoptotic process intracellular signal transduction positive regulation of protein kinase activity response to stress protein autophosphorylation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline