    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • FLNA/FLNB Antibodies
          Invitrogen
          Phospho-FLNA/FLNB (Ser2152, Ser2107) Recombinant Rabbit Monoclonal Antibody (K01_2K92)
          Questions & Answers
          Cite Phospho-FLNA/FLNB (Ser2152, Ser2107) Recombinant Rabbit Monoclonal Antibody (K01_2K92)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          Phospho-FLNA/FLNB (Ser2152, Ser2107) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-FLNA/FLNB (Ser2152, Ser2107) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Phospho-FLNA/FLNB (Ser2152, Ser2107) Antibody (MA567869) in ICC/IF
          Immunocytochemical analysis of Phospho-FLNA/FLNB (Ser2152, Ser2107)(green) in A549 using Phospho-FLNA/FLNB (Ser2152, Ser2107) Recombinant Monoclonal Antibody (Product # MA567869) and DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-FLNA/FLNB (Ser2152, Ser2107) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-FLNA/FLNB (Ser2152, Ser2107) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-FLNA/FLNB (Ser2152, Ser2107) Antibody in Western Blot (WB)
          Product Details
          MA567869
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:50-1:100
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          K01_2K92
          Immunogen
          A synthetic phosphopeptide corresponding to residues surrounding Ser2152 of human Filamin A.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A/G
          Storage buffer
          50mM tris glycine, pH 7.4, with 40% glycerol, 150mM NaCl, 0.05% BSA
          Contains
          0.01% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          FLNA and FLNB encode filamin A and filamin B, large actin binding scaffold proteins that crosslink F actin into orthogonal networks and anchor signaling receptors and ion channels to the cytoskeleton. Each filamin forms a dimer through a C terminal domain and contains an N terminal actin binding module followed by multiple immunoglobulin like repeats that provide mechanical flexibility and sites for interaction with membrane proteins, small GTPases, kinases, and transcriptional regulators. FLNA is broadly expressed and crucial for cell migration, mechanosensing, platelet function, and neuronal development, whereas FLNB has prominent roles in skeletal patterning, chondrocyte differentiation, and tissue morphogenesis. Pathogenic variants in FLNA cause periventricular nodular heterotopia, cardiovascular anomalies, and platelet disorders, while FLNB mutations lead to skeletal dysplasias such as atelosteogenesis and boomerang dysplasia, highlighting nonredundant functions despite overlapping activities. Filamins serve as mechanotransducers that couple extracellular matrix forces to intracellular signaling and gene expression, and they are studied using actin dynamics assays, interaction proteomics, imaging of cytoskeletal architecture, and genetic perturbation in cellular and animal models.
          Bioinformatics
          Protein Aliases: ABP-278; ABP-280; ABP-280 homolog; actin binding protein 278; actin binding protein 280; Actin-binding protein 280; Actin-binding-like protein; Alpha-filamin; Beta-filamin; Endothelial actin-binding protein; Endothelial actin-binding protein 280); epididymis secretory sperm binding protein; Fh1; Filamin A; filamin A, alpha; Filamin B; filamin B, beta; Filamin homolog 1; filamin, alpha; filamin, beta; Filamin-1; Filamin-3; Filamin-A; Filamin-B; FLN-A; FLN-B; FLN1; FLN3; homolog of ABP-280; Larsen syndrome 1 (autosomal dominant); Non-muscle filamin; Thyroid autoantigen; Truncated ABP; Truncated actin-binding protein; unnamed protein product
          View more View less
          Gene Aliases: ABP-278; ABP-280; ABPX; AOI; CSBS; CVD1; FGS2; FH1; FLN; FLN-A; FLN-B; FLN1; FLN1L; FLN3; FLNA; FLNB; FMD; LRS1; MNS; NHBP; OPD; OPD1; OPD2; RGD1560614; SCT; TABP; TAP; XLVD; XMVD
          View more View less
          UniProt ID: (Human) P21333, (Human) B2ZZ83
          View more View less
          Entrez Gene ID: (Human) 2316, (Human) 2317, (Rat) 293860, (Rat) 306204
          View more View less
          Function(s)
          G-protein coupled receptor binding RNA binding actin binding protein binding potassium channel regulator activity kinase binding small GTPase binding Fc-gamma receptor I complex binding protein homodimerization activity ion channel binding macromolecular complex binding cadherin binding SMAD binding actin filament binding GTPase binding DNA-binding transcription factor binding protein sequestering activity identical protein binding glycoprotein binding signal transducer activity protein kinase C binding transcription factor binding Rho GTPase binding Ral GTPase binding mu-type opioid receptor binding protein complex binding poly(A) RNA binding Rac GTPase binding molecular_function
          Process(es)
          epithelial to mesenchymal transition adenylate cyclase-inhibiting dopamine receptor signaling pathway biological_process actin cytoskeleton organization actin cytoskeleton reorganization protein localization to cell surface negative regulation of protein catabolic process cilium assembly mRNA transcription from RNA polymerase II promoter positive regulation of transcription factor import into nucleus receptor clustering positive regulation of I-kappaB kinase/NF-kappaB signaling negative regulation of sequence-specific DNA binding transcription factor activity early endosome to late endosome transport establishment of protein localization protein stabilization cytoplasmic sequestering of protein actin crosslink formation platelet aggregation mitotic spindle assembly keratinocyte development epithelial cell morphogenesis skeletal muscle tissue development cell differentiation blood coagulation, intrinsic pathway positive regulation of platelet activation negative regulation of transcription from RNA polymerase I promoter formation of radial glial scaffolds cerebral cortex development cell projection organization regulation of cell migration positive regulation of actin filament bundle assembly megakaryocyte development positive regulation of protein import into nucleus negative regulation of apoptotic process wound healing, spreading of cells release of sequestered calcium ion into cytosol cilium morphogenesis semaphorin-plexin signaling pathway protein localization to plasma membrane tubulin deacetylation establishment of Sertoli cell barrier positive regulation of substrate adhesion-dependent cell spreading positive regulation of potassium ion transmembrane transport protein localization to bicellular tight junction regulation of membrane repolarization during atrial cardiac muscle cell action potential regulation of membrane repolarization during cardiac muscle cell action potential positive regulation of neural precursor cell proliferation positive regulation of integrin-mediated signaling pathway positive regulation of neuron migration signal transduction muscle organ development cellular response to interferon-gamma
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline