    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NRF1 Antibodies
          Invitrogen
          NRF1 Recombinant Rabbit Monoclonal Antibody (HL3307)
          View all (34) NRF1 antibodies
          Questions & Answers
          Cite NRF1 Recombinant Rabbit Monoclonal Antibody (HL3307)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          NRF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          NRF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          NRF1 Antibody (MA568160) in IHC (P)
          Immunohistochemistry analysis of NRF1 in paraffin-embedded mouse tissues. Samples were incubated with NRF1 recombinant monoclonal antibody (Product # MA568160) using a dilution of 1:200. Antigen Retrieval: Citrate buffer, pH 60, 15 min. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          NRF1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NRF1 Antibody in Western Blot (WB)
          NRF1 Antibody in Western Blot (WB)
          Product Details
          MA568160
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:1,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100-1:1,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Zebrafish
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          HL3307
          Immunogen
          Recombinant fragment of human NRF1.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          no preservative
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a protein that homodimerizes and functions as a transcription factor which activates the expression of some key metabolic genes regulating cellular growth and nuclear genes required for respiration, heme biosynthesis, and mitochondrial DNA transcription and replication. The protein has also been associated with the regulation of neurite outgrowth. Alternate transcriptional splice variants, which encode the same protein, have been characterized. Additional variants encoding different protein isoforms have been described but they have not been fully characterized. Confusion has occurred in bibliographic databases due to the shared symbol of NRF1 for this gene and for "nuclear factor (erythroid-derived 2)-like 1" which has an official symbol of NFE2L1.
          Bioinformatics
          Protein Aliases: Alpha palindromic-binding protein; Alpha-pal; EWG; Not really finished protein; nrf; NRF-1; nrl; Nuclear respiratory factor 1; unnamed protein product
          View more View less
          Gene Aliases: ALPHA-PAL; C87038; D6Ertd415e; NRF1; SC:BZ3C13.1; SC:bZ3C13.2; ube2h
          View more View less
          UniProt ID: (Human) Q16656, (Mouse) Q9WU00, (Zebrafish) Q90X44
          View more View less
          Entrez Gene ID: (Human) 4899, (Mouse) 18181, (Zebrafish) 64604
          View more View less
          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding protein binding protein homodimerization activity transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding core promoter sequence-specific DNA binding DNA-binding transcription factor
          Process(es)
          regulation of transcription from RNA polymerase II promoter positive regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated transcription from RNA polymerase II promoter mitochondrion organization response to electrical stimulus retina homeostasis neural retina development multicellular organism development response to activity photoreceptor cell maintenance retina development in camera-type eye cellular response to cold
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline