    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GSDME Antibodies
          Invitrogen
          DFNA5 Recombinant Rabbit Monoclonal Antibody (8Y3R1)
          View all (29) GSDME antibodies
          Questions & Answers
          Cite DFNA5 Recombinant Rabbit Monoclonal Antibody (8Y3R1)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          DFNA5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          DFNA5 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          DFNA5 Antibody (MA569502) in ICC/IF
          Immunocytochemistry analysis of DFNA5 on paraffin-embedded NCI-H522 (Doxorubicin-treated) and NCI-H522 (untreated) cell line tissue. Samples were incubated with DFNA5 recombinant monoclonal antibody (Product # MA569502) using a dilution of 1:200 (40x lens). High pressure antigen retrieval performed with 0. 01M Tris-EDTA Buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          DFNA5 Antibody in Immunocytochemistry (ICC/IF)
          DFNA5 Antibody in Immunocytochemistry (ICC/IF)
          DFNA5 Antibody in Western Blot (WB)
          Product Details
          MA569502
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200-1:800
          -
          Immunocytochemistry (ICC/IF)
          1: 200
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          8Y3R1
          Immunogen
          Synthetic peptide.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.65 mg/mL
          Amount
          165 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3., with 50% glycerol, 0.05% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          GSDME (Gasdermin E) is a member of the gasdermin family of pore-forming proteins that regulate programmed cell death and inflammatory signaling. Encoded by the GSDME gene, the full-length protein contains an N-terminal pore-forming domain and a C-terminal autoinhibitory domain. Under apoptotic conditions, activation of caspase-3 leads to proteolytic cleavage of GSDME, releasing the N-terminal fragment, which oligomerizes and inserts into the plasma membrane to form pores. This process can convert non-lytic apoptosis into a lytic, inflammatory form of cell death resembling pyroptosis, resulting in membrane rupture and release of intracellular contents. GSDME has been implicated in cancer biology, chemotherapy response, and tissue injury, and was originally identified in association with autosomal dominant hearing loss (historically referred to as DFNA5). Detection of cleaved GSDME is commonly used as a marker of caspase-3-mediated pore formation and inflammatory cell death.
          Bioinformatics
          Protein Aliases: DFNA5; DFNA5, deafness associated tumor suppressor; expression inversely correlated with estrogen receptor expression in breast cancer; Gasdermin E; Gasdermin-E; ICERE-1; Inversely correlated with estrogen receptor expression 1; Non-syndromic hearing impairment protein 5; nonsyndromic hearing impairment protein; unnamed protein product
          View more View less
          Gene Aliases: DFNA5; GSDME; ICERE-1; ICERE1
          View more View less
          UniProt ID: (Human) O60443
          View more View less
          Entrez Gene ID: (Human) 1687
          View more View less
          Function(s)
          protein binding phosphatidylinositol-4,5-bisphosphate binding wide pore channel activity cardiolipin binding
          Process(es)
          positive regulation of immune response to tumor cell sensory perception of sound negative regulation of cell proliferation programmed cell death auditory receptor cell differentiation positive regulation of MAPK cascade transmembrane transport pyroptosis cellular response to tumor necrosis factor cellular response to virus granzyme-mediated programmed cell death signaling pathway pyroptotic cell death positive regulation of intrinsic apoptotic signaling pathway
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline