    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MFGE8 (Lactadherin) Antibodies
          Invitrogen
          MFGE8 Recombinant Rabbit Monoclonal Antibody (7U4L10)
          View all (25) MFGE8 (Lactadherin) antibodies
          Questions & Answers
          Cite MFGE8 Recombinant Rabbit Monoclonal Antibody (7U4L10)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          MFGE8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          MFGE8 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          MFGE8 Antibody (MA569580) in ICC/IF
          Immunocytochemical analysis of MFGE8 in U-2 OS cells. Samples were incubated with MFGE8 recombinant monoclonal antibody (Product # MA569580) using a dilution of 1:200 (40x lens). Followed by a further incubation with Cy3 Goat Anti-Rabbit IgG (H+L) using a dilution of 1:500 (Red). The cells were counterstained with alpha-tubulin Mouse mAb using dilution of 1:400. Followed by incubation with 488-conjugated Goat Anti-Mouse IgG (H+L) using a dilution of 1:500 (Green). DAPI was used for nuclear staining (Blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          MFGE8 Antibody in Immunocytochemistry (ICC/IF)
          MFGE8 Antibody in Immunocytochemistry (ICC/IF)
          MFGE8 Antibody in Immunocytochemistry (ICC/IF)
          MFGE8 Antibody in Western Blot (WB)
          Product Details
          MA569580
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:100,000-1:600,000
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:800
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          7U4L10
          Immunogen
          Recombinant protein (or fragment).
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          2.04 mg/mL
          Amount
          204 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3., with 50% glycerol
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a preproprotein that is proteolytically processed to form multiple protein products. The major encoded protein product, lactadherin, is a membrane glycoprotein that promotes phagocytosis of apoptotic cells. This protein has also been implicated in wound healing, autoimmune disease, and cancer. Lactadherin can be further processed to form a smaller cleavage product, medin, which comprises the major protein component of aortic medial amyloid (AMA). Alternative splicing results in multiple transcript variants.
          Bioinformatics
          Protein Aliases: AGS; Breast epithelial antigen BA46; EGF/factor VIII; HMFG; Lactadherin; medin; MFG-E8; MFGM; milk fat globule glycoprotein; milk fat globule membrane; milk fat globule membrane protein; milk fat globule protein; Milk fat globule-EGF factor 8; milk fat globule-EGF factor 8 protein; MP47; O-acetyl disialoganglioside synthase; O-acetyl GD3 ganglioside synthase; O-acetyltransferase milk fat globule membrane protein; O-acetyltransferase Milk fat glolbule membrane protein; SED1; sperm associated antigen 10; sperm surface protein hP47; Sperm surface protein SP47; unnamed protein product
          View more View less
          Gene Aliases: AA408458; AGS; AI325141; BA46; EDIL1; HMFG; hP47; HsT19888; lactadherin; MFG-E8; MFGE8; MFGM; MFGME8; MFGMP-E8; MP47; OAcGD3S; P47; SED1; SPAG10
          View more View less
          UniProt ID: (Human) Q08431, (Mouse) P21956, (Rat) P70490
          View more View less
          Entrez Gene ID: (Human) 4240, (Mouse) 17304, (Rat) 25277
          View more View less
          Function(s)
          phosphatidylserine binding integrin binding extracellular matrix structural constituent phosphatidylethanolamine binding oxidoreductase
          Process(es)
          angiogenesis cell adhesion single fertilization apoptotic cell clearance phagocytosis, recognition phagocytosis, engulfment positive regulation of phagocytosis positive regulation of apoptotic cell clearance positive regulation of cell proliferation response to estrogen
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline