    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • METTL3 Antibodies
          Invitrogen
          METTL3 Recombinant Rabbit Monoclonal Antibody (6G8P7)
          View all (28) METTL3 antibodies
          Questions & Answers
          Cite METTL3 Recombinant Rabbit Monoclonal Antibody (6G8P7)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (11)
          METTL3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          METTL3 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 11
          METTL3 Antibody (MA569586) in ICC/IF
          Immunocytochemical analysis of METTL3 in Neuro-2a cells. Samples were incubated with METTL3 recombinant monoclonal antibody (Product # MA569586) using a dilution of 1:200 (100x lens). Additional incubation with Cy3-conjugated Goat anti-Rabbit IgG (H+L) using a dilution of 1:500 (Red). The cells were counterstained with alpha-Tubulin Mouse mAb using a dilution of 1:400. Followed by incubation with 488-conjugated Goat Anti-Mouse IgG (H+L) Ab using a dilution of 1:500 (Green). DAPI was used for nuclear staining (Blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          METTL3 Antibody in Immunocytochemistry (ICC/IF)
          METTL3 Antibody in Immunocytochemistry (ICC/IF)
          METTL3 Antibody in Immunocytochemistry (ICC/IF)
          METTL3 Antibody in Immunocytochemistry (ICC/IF)
          METTL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          METTL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          METTL3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          METTL3 Antibody in Western Blot (WB)
          METTL3 Antibody in Western Blot (WB)
          METTL3 Antibody in Western Blot (WB)
          METTL3 Antibody in ChIP Assay (ChIP)
          Product Details
          MA569586
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000-1:10,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:800-1:3,200
          -
          Immunocytochemistry (ICC/IF)
          1:200-1:400
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Immunoprecipitation (IP)
          0.5-4 µg
          -
          ChIP assay (ChIP)
          5 µg
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          6G8P7
          Immunogen
          Recombinant protein (or fragment).
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.3 mg/mL
          Amount
          130 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3., with 50% glycerol, 0.05% BSA
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          METTL3 is the 70 kDa subunit of MT-A which is part of N6-adenosine-methyltransferase. This enzyme is involved in the posttranscriptional methylation of internal adenosine residues in eukaryotic mRNAs, forming N6-methyladenosine.
          Bioinformatics
          Protein Aliases: adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; hMETTL3; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; Methyltransferase-like protein 3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MT-A70; N(6)-adenosine-methyltransferase 70 kDa subunit; N(6)-adenosine-methyltransferase catalytic subunit METTL3; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase subunit METTL3; unnamed protein product
          View more View less
          Gene Aliases: 2310024F18Rik; hMETTL3; IME4; M6A; METTL3; MT-A70; MTA70; Spo8
          View more View less
          UniProt ID: (Human) Q86U44, (Mouse) Q8C3P7
          View more View less
          Entrez Gene ID: (Human) 56339, (Mouse) 56335, (Rat) 361035
          View more View less
          Function(s)
          mRNA (N6-adenosine)-methyltransferase activity RNA binding mRNA binding protein binding methyltransferase activity RNA methyltransferase activity transferase activity protein heterodimerization activity S-adenosyl-L-methionine binding mRNA (2'-O-methyladenosine-N6-)-methyltransferase activity RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein
          Process(es)
          mRNA splicing, via spliceosome RNA methylation immune system process mRNA processing mRNA export from nucleus cellular response to DNA damage stimulus spermatogenesis circadian rhythm dosage compensation by inactivation of X chromosome mRNA modification stem cell population maintenance forebrain radial glial cell differentiation cell differentiation primary miRNA processing methylation cellular response to UV gliogenesis innate immune response regulation of T cell differentiation positive regulation of translation negative regulation of Notch signaling pathway oogenesis rhythmic process regulation of meiotic cell cycle negative regulation of type I interferon-mediated signaling pathway mRNA destabilization endothelial to hematopoietic transition regulation of hematopoietic stem cell differentiation positive regulation of cap-independent translational initiation nucleobase-containing compound metabolic process adenosine to inosine editing mRNA methylation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline