    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • HSD17B2 Antibodies
          Invitrogen
          HSD17B2 Recombinant Rabbit Monoclonal Antibody (10Y8D4)
          View all (18) HSD17B2 antibodies
          Questions & Answers
          Cite HSD17B2 Recombinant Rabbit Monoclonal Antibody (10Y8D4)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (6)
          HSD17B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          HSD17B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 6
          HSD17B2 Antibody (MA569656) in IHC (P)
          Immunohistochemistry analysis of HSD17B2 on paraffin-embedded Mouse liver tissue. Samples were incubated with HSD17B2 recombinant monoclonal antibody (Product # MA569656) using a dilution of 1:1,000 (40x lens). High pressure antigen retrieval performed with 0. 01 M Tris-EDTA Buffer (pH 9. 0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HSD17B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSD17B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSD17B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSD17B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSD17B2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          HSD17B2 Antibody in Western Blot (WB)
          Product Details
          MA569656
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:13,000-1:78,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:700-1:7,000
          -
          ELISA (ELISA)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          10Y8D4
          Immunogen
          Synthetic peptide.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1.37 mg/mL
          Amount
          137 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.3., with 50% glycerol
          Contains
          0.09% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          HSD17B2 is capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. HSD17B2 also has 20-alpha-HSD activity. HSD17B2 uses NADH while EDH17B3 uses NADPH.
          Bioinformatics
          Protein Aliases: 1.1.1.239; 17 beta HSD 2; 17 HSD type 2; 17-beta hydroxysteroid dehydrogenase type 2; 17-beta-HSD 2; 17-beta-hydroxysteroid dehydrogenase 2; 17-beta-hydroxysteroid dehydrogenase type 2; 20 alpha HSD; 20 alpha-hydroxysteroid dehydrogenase; 20-alpha-HSD; E2DH; Estradiol 17-beta-dehydrogenase 2; Microsomal 17-beta-hydroxysteroid dehydrogenase; Short chain dehydrogenase/reductase family 9C member 2; tes; Testosterone 17-beta-dehydrogenase; unnamed protein product
          View more View less
          Gene Aliases: AI194836; AI194967; AI255511; EDH17B2; HSD17; HSD17B2; SDR9C2
          View more View less
          UniProt ID: (Human) P37059, (Mouse) P51658, (Rat) Q62730
          View more View less
          Entrez Gene ID: (Human) 3294, (Mouse) 15486, (Rat) 79243
          View more View less
          Function(s)
          estradiol 17-beta-dehydrogenase activity protein binding oxidoreductase activity oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor 17-alpha,20-alpha-dihydroxypregn-4-en-3-one dehydrogenase activity testosterone dehydrogenase (NAD+) activity dehydrogenase oxidoreductase metabolite interconversion enzyme
          Process(es)
          in utero embryonic development placenta development lipid metabolic process steroid biosynthetic process estrogen biosynthetic process steroid metabolic process androgen metabolic process response to retinoic acid androgen biosynthetic process metabolic process oxidation-reduction process bone development cellular response to metal ion
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline