    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • ULBP3 Antibodies
          Invitrogen
          ULBP3 Recombinant Rabbit Monoclonal Antibody (PSH13-03)
          View all (8) ULBP3 antibodies
          Questions & Answers
          Cite ULBP3 Recombinant Rabbit Monoclonal Antibody (PSH13-03)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (5)
          ULBP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ULBP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          ULBP3 Antibody (MA570004) in IHC (P)
          Immunohistochemistry analysis of ULBP3 in paraffin-embedded human placenta tissue. Samples were incubated with ULBP3 monoclonal antibody (Product # MA570004) using a dilution of 1:200 for 1 hour at room temperature. The section was pre-treated using heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0) for 20 minutes. The tissues were blocked in 1% BSA for 20 minutes at room temperature, washed wit... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          ULBP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ULBP3 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ULBP3 Antibody in Western Blot (WB)
          ULBP3 Antibody in Western Blot (WB)
          ULBP3 Antibody in Immunoprecipitation (IP)
          ULBP3 Recombinant Rabbit Monoclonal Antibody (PSH13-03)
          Product Details
          MA570004
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:200
          -
          Immunoprecipitation (IP)
          1-2 µg
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          PSH13-03
          Immunogen
          Recombinant protein within human ULBP3 aa 21-220.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.1% BSA, 40% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Positive control: MDA-MB-231 cell lysate, MCF7 cell lysate, SW480 cell lysate, HeLa cell lysate, human placenta tissue, human stomach tissue.
          Subcellular location: Cell membrane.
          Target Information
          UL16-binding protein 3 (ULBP3) is an extended member of the MHC class I superfamily. It does not contain the alpha 3 domain and lacks a transmembrane domain. ULBP3 binds and activates the KLRK1/NKG2D receptor and mediates natural killer cell cytotoxicity.
          Bioinformatics
          Protein Aliases: ALCAN-gamma; N2DL-3; NKG2D ligand 3; NKG2D ligand3; Retinoic acid early transcript 1N; UL16-binding protein 3; ULBP; ULBP 3; unnamed protein product
          View more View less
          Gene Aliases: N2DL-3; N2DL3; NKG2DL3; RAET1N; ULBP3
          View more View less
          UniProt ID: (Human) Q9BZM4
          View more View less
          Entrez Gene ID: (Human) 79465
          View more View less
          Function(s)
          protein binding natural killer cell lectin-like receptor binding
          Process(es)
          positive regulation of T cell mediated cytotoxicity immune system process antigen processing and presentation of endogenous peptide antigen via MHC class Ib antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-independent immune response natural killer cell activation natural killer cell mediated cytotoxicity
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline