    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Cystatin F Antibodies
          Invitrogen
          Cystatin F Recombinant Rabbit Monoclonal Antibody (PSH09-00)
          View all (10) Cystatin F antibodies
          Questions & Answers
          Cite Cystatin F Recombinant Rabbit Monoclonal Antibody (PSH09-00)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          Cystatin F Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Cystatin F Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          Cystatin F Antibody (MA570069) in IHC (F)
          Immunohistochemistry analysis of Cystatin F in frozen mouse brain tissue (negative). Samples were incubated with Cystatin F monoclonal antibody (Product # MA570069) using a dilution of 1:100 overnight at 4°C, washed with PBS followed by Goat Anti-Rabbit IgG H&L (iFluor™ 488) at a dilution of 1:1,000. The section was pre-treated using heat mediated antigen retrieval with sodium citrate buffer (pH 6.0) for ab... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Cystatin F Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cystatin F Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Cystatin F Antibody in Western Blot (WB)
          Cystatin F Recombinant Rabbit Monoclonal Antibody (PSH09-00)
          Product Details
          MA570069
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunohistochemistry (Frozen) (IHC (F))
          1:100
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          PSH09-00
          Immunogen
          Recombinant protein within human Cystatin F aa 1-145.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.1% BSA, 40% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Positive control: U-937 cell lysate, mouse spleen tissue.
          Subcellular location: Secreted, Cytoplasm.
          Target Information
          The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. This gene encodes a glycosylated cysteine protease inhibitor with a putative role in immune regulation through inhibition of a unique target in the hematopoietic system. Expression of the protein has been observed in various human cancer cell lines established from malignant tumors.
          Bioinformatics
          Protein Aliases: CMAP; CST7; cystatin 7; Cystatin-7; Cystatin-F; Cystatin-like metastasis-associated protein; Leukocystatin
          View more View less
          Gene Aliases: CMAP; CST7
          View more View less
          UniProt ID: (Human) O76096, (Mouse) O89098
          View more View less
          Entrez Gene ID: (Human) 8530, (Mouse) 13011
          View more View less
          Function(s)
          protease binding endopeptidase inhibitor activity cysteine-type endopeptidase inhibitor activity protein binding peptidase inhibitor activity protein homodimerization activity
          Process(es)
          negative regulation of peptidase activity regulation of antigen processing and presentation immune response positive regulation of myelination negative regulation of microglial cell activation lysosomal protein catabolic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline