    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • B7-H6 Antibodies
          Invitrogen
          B7-H6 Recombinant Rabbit Monoclonal Antibody (PSH10-89)
          View all (18) B7-H6 antibodies
          Questions & Answers
          Cite B7-H6 Recombinant Rabbit Monoclonal Antibody (PSH10-89)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          B7-H6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          B7-H6 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          B7-H6 Antibody (MA570139) in ICC/IF
          Immunocytochemistry analysis of B7-H6 in K-562 (positive) and Daudi (negative). Samples were incubated with B7-H6 monoclonal antibody (Product # MA570139) using a dilution of 1:100 in 1% BSA in PBST overnight at 4°C followed by Goat Anti-Rabbit IgG H&L (iFluor™ 488) at a dilution of 1:1,000. Cells were fixed in 4% paraformaldehyde for 15 minutes at room temperature, permeabilized with 0.1% Triton X-100 in PBS for 15 minutes at room temperature, then blocked with 1% BSA in 10% negative goat serum for 1 hour at room temperature. PBS instead ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          B7-H6 Antibody in Immunocytochemistry (ICC/IF)
          B7-H6 Antibody in Western Blot (WB)
          B7-H6 Antibody in Flow Cytometry (Flow)
          B7-H6 Antibody in Immunoprecipitation (IP)
          B7-H6 Recombinant Rabbit Monoclonal Antibody (PSH10-89)
          Product Details
          MA570139
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:2,000
          -
          Immunocytochemistry (ICC/IF)
          1:100
          -
          Flow Cytometry (Flow)
          1:5,000
          -
          Immunoprecipitation (IP)
          1-2 µg
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293 cells
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          PSH10-89
          Immunogen
          Recombinant protein within human B7-H6 aa 1-262.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4, with 0.1% BSA, 40% glycerol
          Contains
          0.05% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Positive control: K-562 cell lysate, HepG2 cell lysate, HeLa cell lysate.
          Subcellular location: Cell membrane.
          Target Information
          B7-H6 (NCR3LG1) is a transmembrane endogenous ligand expressed on the surfaces of tumor cells. B7-H6 binds with NKp30 present on NK cells. Binding of B7-H6 to NKp30 causes the ligation of NKp30 subsequently inducing NK cell activation and target cell cytolysis. B7-H6 is not constitutively expressed on normal tissue. Whether it can be expressed endogenously under certain conditions is under investigation. B7-H6 has been detected on circulating pro-inflammatory CD14(+)CD16(+) monocytes in some patients with sepsis.
          Bioinformatics
          Protein Aliases: B7 homolog 6; B7-H6; DKFZp686I21167; Natural cytotoxicity triggering receptor 3 ligand 1; putative Ig-like domain-containing protein DKFZp686O24166/DKFZp686I21167
          View more View less
          Gene Aliases: B7-H6; B7H6; DKFZp686O24166; NCR3LG1
          View more View less
          UniProt ID: (Human) Q68D85
          View more View less
          Entrez Gene ID: (Human) 374383
          View more View less
          Function(s)
          protein binding receptor agonist activity
          Process(es)
          signal transduction NK T cell activation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline