    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GYS1 Antibodies
          Invitrogen
          GYS1 Recombinant Rabbit Monoclonal Antibody (25GB2105)
          View all (32) GYS1 antibodies
          Questions & Answers
          Cite GYS1 Recombinant Rabbit Monoclonal Antibody (25GB2105)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          GYS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GYS1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          GYS1 Antibody (MA570379) in WB
          Western blot analysis of GYS1. Samples (HT-1080, HepG2, HeLa, H9C2, NIH/3T3) were incubated with GYS1 monoclonal antibody (Product # MA570379) at a dilution of 1:20,000, followed by HRP-conjugated goat anti-rabbit secondary antibody (1:20,000) respectively. Image was developed using NaQECL Substrate Kit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GYS1 Antibody in Western Blot (WB)
          GYS1 Antibody in Western Blot (WB)
          GYS1 Antibody in Flow Cytometry (Flow)
          GYS1 Antibody in Flow Cytometry (Flow)
          GYS1 Recombinant Rabbit Monoclonal Antibody (25GB2105)
          Product Details
          MA570379
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:20,000
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          Flow Cytometry (Flow)
          1:2,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          25GB2105
          Immunogen
          A synthesized peptide derived from human (700-737) Glycogen synthase.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Target Information
          Glycogen Synthase 1 (GYS1) catalyzes the addition of glucose monomers to the growing glycogen molecule through the formation of alpha-1,4-glycoside linkages. Mutations in this gene are associated with muscle glycogen storage disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
          Bioinformatics
          Protein Aliases: EC 2.4.1.11; Glycogen [starch] synthase, muscle; glycogen synthase; Glycogen synthase 1; glycogen synthase 1 (muscle); glycogen synthase 3, brain; unnamed protein product
          View more View less
          Gene Aliases: GSY; GYS; GYS1; Gys3; MGS
          View more View less
          UniProt ID: (Human) P13807, (Rat) A2RRU1, (Mouse) Q9Z1E4
          View more View less
          Entrez Gene ID: (Human) 2997, (Rat) 690987, (Mouse) 14936
          View more View less
          Function(s)
          catalytic activity glycogen (starch) synthase activity protein binding glucose binding transferase activity transferase activity, transferring glycosyl groups glycogen synthase activity, transferring glucose-1-phosphate protein kinase binding
          Process(es)
          glycogen metabolic process glycogen biosynthetic process heart development metabolic process
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline