    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TRAP alpha Antibodies
          Invitrogen
          TRAP alpha Recombinant Rabbit Monoclonal Antibody (25GB1955)
          View all (29) TRAP alpha antibodies
          Questions & Answers
          Cite TRAP alpha Recombinant Rabbit Monoclonal Antibody (25GB1955)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          TRAP alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          TRAP alpha Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          TRAP alpha Antibody (MA570533) in WB
          Western blot analysis of TRAP alpha. Samples (HT-1080, HepG2, HeLa, HAP-1, H9C2, C2C12) were incubated with TRAP alpha monoclonal antibody (Product # MA570533) at a dilution of 1:5,000, followed by HRP-conjugated goat anti-rabbit secondary antibody (1:20,000) respectively. Image was developed using NaQECL Substrate Kit. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRAP alpha Antibody in Western Blot (WB)
          TRAP alpha Antibody in Western Blot (WB)
          TRAP alpha Antibody in Flow Cytometry (Flow)
          TRAP alpha Antibody in Flow Cytometry (Flow)
          TRAP alpha Recombinant Rabbit Monoclonal Antibody (25GB1955)
          Product Details
          MA570533
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:5,000
          -
          Flow Cytometry (Flow)
          1:2,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Expression System
          HEK293F
          Class
          Recombinant Monoclonal
          Type
          Antibody
          Clone
          25GB1955
          Immunogen
          A synthesized peptide derived from human (1-286) TRAP alpha.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Dry ice (international)
          Target Information
          The signal sequence receptor (SSR) is a glycosylated endoplasmic reticulum (ER) membrane receptor associated with protein translocation across the ER membrane. The SSR consists of 2 subunits, a 34-kD glycoprotein encoded by this gene and a 22-kD glycoprotein. This gene generates several mRNA species as a result of complex alternative polyadenylation. This gene is unusual in that it utilizes arrays of polyA signal sequences that are exclusively non-canonical.
          Bioinformatics
          Protein Aliases: DKFZp781N23103; liver regeneration-related protein LRRG137; Signal sequence receptor subunit alpha; signal sequence receptor, alpha; SSR alpha subunit; SSR-alpha; SSR1; translocon-associated protein alpha; translocon-associated protein alpha subunit; Translocon-associated protein subunit alpha; TRAP alpha; TRAP-alpha; unnamed protein product
          View more View less
          Gene Aliases: 2510001K09Rik; 6330400D04; Ac2-238; AI159733; AI452176; PSEC0262; SSR; SSR1; TRAPA
          View more View less
          UniProt ID: (Human) P43307, (Mouse) Q9CY50
          View more View less
          Entrez Gene ID: (Human) 6745, (Mouse) 107513, (Rat) 361233
          View more View less
          Function(s)
          protein binding membrane traffic protein
          Process(es)
          cotranslational protein targeting to membrane positive regulation of cell proliferation
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline