    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD49d (Integrin alpha 4) Antibodies
          Invitrogen
          ITGA4 Monoclonal Antibody (9F10)
          View all (74) CD49d (Integrin alpha 4) antibodies
          Questions & Answers
          Cite ITGA4 Monoclonal Antibody (9F10)
          Additional Information:
          {{banner.description}}
          Product Details
          MA574482
          Applications
          Tested Dilution
          Publications
          Immunohistochemistry (Frozen) (IHC (F))
          5-10 µg/mL
          -
          Flow Cytometry (Flow)
          1 µg/mL
          -
          Product Specifications
          Species Reactivity
          Bovine, Dog, Horse, Cat, Human, Non-human primate, Sheep
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          9F10
          Immunogen
          The immunogen information is proprietary.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • APC-Cyanine7
          • FITC
          • Functional Grade
          • PE 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4
          Contains
          15mM sodium azide
          Storage conditions
          4°C, do not freeze
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          The integrin alpha 4 (also known as CD49d and ITGA4) belongs to the integrin alpha chain family of proteins. Integrins are heterodimeric integral membrane proteins composed of an alpha and beta chains. Alpha4 chain associates with either beta1 or beta7 chain. It has been demonstrated that the putative ligand-binding sites of both integrin alpha4beta1 and alpha4beta7 is located on the alpha4 chain. These ligands included Madcam, VCAM, and fibronectin. Madcam is known as the principal ligand for integrin alpha4beta7. Recently it was also demonstrated that HIV-1 envelope can mimic Madcam by binding to and signaling though integrin alpha4beta7, the gut mucosal homing receptor for peripheral T cells.
          Bioinformatics
          Protein Aliases: CD49 antigen-like family member D; CD49d; Integrin alpha-4; Integrin alpha-4-like protein; Integrin alpha-IV; integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor); ITGA-IV; VLA-4 subunit alpha
          View more View less
          Gene Aliases: CD49D; ITGA4
          View more View less
          Entrez Gene ID: (Sheep) 443499, (Horse) 100067886, (Dog) 488429, (Bovine) 282882
          View more View less
          Function(s)
          integrin
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline