    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • EVI2B Antibodies
          Invitrogen
          CD361 Monoclonal Antibody (MEM-216), APC
          View all (8) EVI2B antibodies
          Questions & Answers
          Cite CD361 Monoclonal Antibody (MEM-216), APC
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          CD361 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD361 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD361 Antibody (MA574646) in Flow
          Flow cytometry analysis of CD361 in human peripheral blood. Samples were incubated with monoclonal CD361 (Product # MA574646), followed by APC. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD361 Antibody in Flow Cytometry (Flow)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          CD361 Monoclonal Antibody (MEM-216), APC
          Product Details
          MA574646
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          10 µL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MEM-216
          Immunogen
          Raji cells.
          3D Epitope / Immunogen
          Conjugate
          APC APC APC
          • Unconjugated
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          651/660 nm View spectra spectra
          Form
          Liquid
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD361, also known as EVI2B (Ecotropic Viral Integration site 2B) or EVDB, is a poorly characterized type I transmembrane protein, expressed from one of three genes embedded in intron 27b of the neurofibromatosis type 1 (NF1) gene. The DNA strand that is transcribed to produce CD361 is the complementary one to the strand encoding NF1. Murine homolog to human CD361 is associated with ecotropic viral insertions, which have been implicated in the expression of murine myeloid leukemias. CD361 has been also reported to be involved in melanocyte and keratinocyte differentiation. However, it is expressed mainly in peripheral blood and bone marrow.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: ecotropic viral integration site 2B protein homolog; EVI-2B; unnamed protein product
          View more View less
          Gene Aliases: CD361; D17S376; EVDB
          View more View less
          UniProt ID: (Human) P34910
          View more View less
          Entrez Gene ID: (Human) 2124
          View more View less
          Function(s)
          protein binding
          Process(es)
          positive regulation of granulocyte differentiation negative regulation of apoptotic process positive regulation of neutrophil differentiation regulation of cell cycle myeloid cell development regulation of stem cell division
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch