    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD138 (Syndecan-1) Antibodies
          Invitrogen
          CD138 Monoclonal Antibody (MI15), APC
          View all (105) CD138 (Syndecan-1) antibodies
          Questions & Answers
          Cite CD138 Monoclonal Antibody (MI15), APC
          Additional Information:
          {{banner.description}}
          CD138 Monoclonal Antibody (MI15), APC
          Product Details
          MA575174
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          10 µL/10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human, Non-human primate
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          MI15
          Immunogen
          A mixture of U266 and XG-1 human myeloma cell lines.
          3D Epitope / Immunogen
          Conjugate
          APC APC APC
          • Unconjugated
          • FITC
          • Pacific Orange
          • PE 
          • PerCP 
          • Request custom conjugation
          Excitation/Emission Max
          651/660 nm View spectra spectra
          Form
          Liquid
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CD antigen 138; CD138; sCD138; soluble CD138; SYND1; Syndecan-1; Syndecan1
          View more View less
          Gene Aliases: SDC; SDC1
          View more View less
          Entrez Gene ID: (Human) 6382
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-dd86746ff-hszvs:80/100.66.78.131:80.
          git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.53.1-Offline