    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Actin Muscle Antibodies
          Invitrogen
          Actin (muscle) Monoclonal Antibody (HHF35), Alexa Fluor™ 488
          View all (19) Actin Muscle antibodies
          Questions & Answers
          Cite Actin (muscle) Monoclonal Antibody (HHF35), Alexa Fluor™ 488
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Actin (muscle) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Actin (muscle) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Actin (muscle) Antibody (MA575276) in WB
          Western blot analysis of Actin (muscle) in murine skeletal muscle and murine heart (mixed, heated at 100°C, 5 min with reducing SDS-loading buffer). Samples were incubated with monoclonal Actin (muscle) (Product # MA575276) using a dilution of 2 µg/mL, followed by Alexa Fluor 488. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Actin (muscle) Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Actin (muscle) Monoclonal Antibody (HHF35), Alexa Fluor™ 488
          Product Details
          MA575276
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          2 µg/mL
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-dependent
          -
          Immunohistochemistry (Frozen) (IHC (F))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Dog, Chicken, Cat, Human, Mouse, Non-human primate, Rabbit, Rat
          Host/Isotype
          Mouse / IgG1
          Class
          Monoclonal
          Type
          Antibody
          Clone
          HHF35
          Immunogen
          SDS extracted protein fraction of human myocardium.
          3D Epitope / Immunogen
          Conjugate
          Alexa Fluor™ 488 Alexa Fluor™ 488 Alexa Fluor™ 488
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          499/520 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          Size-exclusion chromatography
          Storage buffer
          PBS, pH 7.4, with 0.2% BSA
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          The product encoded by this gene belongs to the actin family of proteins, which are highly conserved proteins that play a role in cell motility, structure and integrity. Alpha, beta and gamma actin isoforms have been identified, with alpha actins being a major constituent of the contractile apparatus, while beta and gamma actins are involved in the regulation of cell motility. This actin is an alpha actin that is found in skeletal muscle. Mutations in this gene cause nemaline myopathy type 3, congenital myopathy with excess of thin myofilaments, congenital myopathy with cores, and congenital myopathy with fiber-type disproportion, diseases that lead to muscle fiber defects.
          Bioinformatics
          Protein Aliases: a-actin; ACTA1; ACTA2; ACTC; ACTG2; actin; actin alpha 1; actin alpha cardiac; actin, alpha 1, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha skeletal muscle; actin, alpha, cardiac 1; actin, alpha, vascular smooth muscle; actin, cytoplasmic 2; actin, cytoplasmic, type 5; actin, gamma 1 propeptide; actin, gamma 2, smooth muscle, enteric; actin, gamma, cytoplasmic 1; actin-like protein; alpha-actin; alpha-actin cardiac; Alpha-actin-1; alpha-actin-2; alpha-actin-3; alpha-cardiac actin; alpha-sma; alpha-smooth muscle actin; alphac-actin; beta-actin FE-3; cardiac actin; cell growth-inhibiting gene 46 protein; cytoplasmic 2; cytoplasmic beta-actin; cytoplasmic type 5; cytoskeletal gamma-actin; epididymis luminal protein 176; gamma 1 propeptide; gamma-2-actin; gamma-actin; gamma-enteric smooth muscle actin; GIG46; nemaline myopathy type 3; RP5-1068B5.2; skeletal muscle alpha-actin; smooth muscle alpha-actin; smooth muscle gamma-actin; type 5; unnamed protein product
          View more View less
          Gene Aliases: 0610041G09Rik; a-SMA; ACT; Act-4; Act4; ACTA; Acta-2; ACTA1; ACTA3; ACTC; Actc-1; ACTE; ACTG; ACTGE; Actl; ACTL3; Acts; ACTSA; ACTSG; Actsk-1; Actvs; alphaSMA; ASD5; ASMA; CFTD; CFTD1; CFTDM; CMD1R; CMH11; CMYO2A; CMYO2B; CMYO2C; CMYP2A; CMYP2B; CMYP2C; DFNA20; DFNA26; E51; GAMMA-ACTIN; HEL-176; LVNC4; MMIHS5; MPFD; NEM1; NEM2; NEM3; SHPM; SMAalpha; SMalphaA; SMDYS; SMGA; VSCM; VSCM1
          View more View less
          UniProt ID: (Human) P68133, (Chicken) P68139, (Rabbit) P68135, (Rabbit) P62740, (Chicken) P08023, (Human) P62736, (Chicken) P68034, (Human) P68032, (Chicken) P53478, (Human) P63261, (Chicken) P63270, (Human) P63267, (Mouse) P68134, (Rat) P68136, (Rat) P62738, (Mouse) P62737, (Rat) P68035, (Mouse) P68033, (Mouse) P63260, (Rat) P63259, (Rat) P63269, (Mouse) P63268
          View more View less
          Entrez Gene ID: (Human) 58, (Chicken) 421534, (Rabbit) 100009506, (Dog) 488984, (Dog) 477587, (Rabbit) 100009271, (Chicken) 423787, (Human) 59, (Cat) 100144392, (Dog) 478250, (Chicken) 423298, (Human) 70, (Rabbit) 100340339, (Chicken) 415296, (Dog) 475923, (Human) 71, (Chicken) 396084, (Human) 72, (Rabbit) 100350318, (Dog) 475792, (Mouse) 11459, (Rat) 29437, (Rat) 81633, (Mouse) 11475, (Rat) 29275, (Mouse) 11464, (Mouse) 11465, (Rat) 287876, (Rat) 25365, (Mouse) 11468
          View more View less
          Function(s)
          structural constituent of cytoskeleton protein binding ATP binding ATPase activity myosin binding ADP binding protein kinase binding microfilament motor activity structural constituent of postsynaptic actin cytoskeleton profilin binding ubiquitin protein ligase binding identical protein binding actin and actin related protein actin or actin-binding cytoskeletal protein cytoskeletal protein
          Process(es)
          muscle contraction positive regulation of gene expression skeletal muscle thin filament assembly skeletal muscle fiber development mesenchyme migration cytoskeleton organization response to virus positive regulation of hepatic stellate cell migration positive regulation of hepatic stellate cell contraction positive regulation of ERK1 and ERK2 cascade cellular response to transforming growth factor beta stimulus juxtaglomerular apparatus development glomerular mesangial cell development positive regulation of hepatic stellate cell activation actin filament organization actin-myosin filament sliding heart contraction actin filament-based movement actomyosin structure organization cardiac myofibril assembly cardiac muscle tissue morphogenesis axonogenesis cell motility postsynaptic actin cytoskeleton organization angiogenesis morphogenesis of a polarized epithelium positive regulation of cell migration maintenance of blood-brain barrier regulation of stress fiber assembly regulation of focal adhesion assembly platelet aggregation positive regulation of wound healing tight junction assembly regulation of transepithelial transport regulation of synaptic vesicle endocytosis protein localization to bicellular tight junction response to ethanol cellular response to interleukin-6 cellular response to acetaldehyde response to mechanical stimulus cellular response to potassium ion skeletal muscle fiber adaptation response to steroid hormone regulation of blood pressure vascular smooth muscle contraction response to xenobiotic stimulus negative regulation of apoptotic process cardiac muscle contraction cytoplasmic actin-based contraction involved in cell motility sarcomere organization cellular response to interferon-gamma response to calcium ion
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-5799cd5976-2rlvt:80/100.66.78.131:80.
          git-commit: e30ab741173a2eb62411af663260592d4bdb9634
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.55.0-2026.07.30-1.0