Search Thermo Fisher Scientific
Reconstitute with 1 ml of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.
For FACS analysis, use 10ul of the suggested working dilution to label 1x10^6 cells in 100ul.
The protein encoded by this gene is an endothelial cell adhesion molecule that interacts preferentially with the leukocyte beta7 integrin LPAM-1 (alpha4beta7), L-selectin, and VLA-4 (alpha4beta1) on myeloid cells to direct leukocytes into mucosal and inflamed tissues. It is a member of the immunoglobulin family and is similar to ICAM1 and VCAM1. At least seven alternatively spliced transcripts encoding different protein isoforms have been found for this gene, but the full-length nature of some variants has not been determined.