Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MFN2 Antibodies

          Invitrogen

          MFN2 Polyclonal Antibody

          1 Reference
          View all (49) MFN2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MFN2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          MFN2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MFN2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 5

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MFN2 Antibody (PA5-79666) in WB

          Western blot was performed using Anti-MFN2 Polyclonal Antibody (Product # PA5-79666) and a 82kDa band corresponding to Mitofusin-2 was observed across cell lines and tissues tested. Additional uncharacterized bands (*) were observed across the panel. Whole cell extracts (30 µg lysate) of A549 (Lane 1), SH-SY5Y (Lane 2), Hep G2 (Lane 3), LNCaP (Lane 4), DU 145 (Lane 5), PC-3 (Lane 6), Mouse spleen (Lane 7), Mouse Kidney (Lane 8) and Mouse Heart (Lane 9)were electrophoresed using NuPAGE™ 10% Bis-Tris Protein Gel (Product # NP0302BOX). Resolv... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MFN2 Antibody in Western Blot (WB)
          MFN2 Antibody in Western Blot (WB)
          MFN2 Antibody in Western Blot (WB)
          MFN2 Antibody in Western Blot (WB)
          MFN2 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          MFN2 Polyclonal Antibody

          Product Details

          PA5-79666

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-0.5 µg/mL
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Hamster

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          E.coli-derived human Mitofusin 2 recombinant protein (Position: V601-R757).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          500 µg/mL once reconstituted

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 5mg BSA

          Contains

          0.05mg sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2746781

          Product Specific Information

          Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 µg/mL.

          Positive Control - WB: HELA whole cell, A549 whole cell, rat brain tissue, rat heart tissue, rat kidney tissue, mouse brain tissue, mouse heart tissue, mouse kidney tissue, mouse small intestine tissue, mouse NIH3T3 whole cell.

          Target Information

          This gene encodes a mitochondrial membrane protein that participates in mitochondrial fusion and contributes to the maintenance and operation of the mitochondrial network. This protein is involved in the regulation of vascular smooth muscle cell proliferation, and it may play a role in the pathophysiology of obesity. Mutations in this gene cause Charcot-Marie-Tooth disease type 2A2, and hereditary motor and sensory neuropathy VI, which are both disorders of the peripheral nervous system. Defects in this gene have also been associated with early-onset stroke. Two transcript variants encoding the same protein have been identified.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CPRP 1; HRP1; HSG protein; hyperplasia suppressor; hypertension related protein 1; hypertension-related protein; MARF; GTP-binding transmembrane protein; MARF; mitochondrial GTP-binding transmembrane protein; MFN 2; mitochondrial assembly regulatory factor; mitochondrial transmembrane GTPase FZO1A; mitofusin-2-like; Mitofusin2; related to proliferation of VSMCs; transmembrane GTPase MFN2; unnamed protein product

          View more View less

          Gene Aliases: CMT2A; CMT2A2; CMT2A2A; CMT2A2B; CPRP1; D630023P19Rik; Fzo; HMSN6A; HSG; MARF; MSL

          View more View less

          UniProt ID: (Human) O95140, (Mouse) Q80U63

          View more View less

          Entrez Gene ID: (Human) 9927, (Rat) 64476, (Mouse) 170731

          View more View less

          Function(s)
          GTPase activity protein binding GTP binding ubiquitin protein ligase binding small GTPase binding identical protein binding proteoglycan binding GTPase binding
          Process(es)
          blastocyst formation mitochondrion organization mitochondrial membrane organization spermatogenesis mitochondrial fusion negative regulation of cell proliferation gonad development male gonad development aerobic respiration response to manganese ion positive regulation of gene expression negative regulation of cardiac muscle cell apoptotic process positive regulation of hydrogen peroxide biosynthetic process positive regulation of mitochondrial membrane potential response to muscle activity response to nutrient levels response to testosterone protein localization to pre-autophagosomal structure negative regulation of cell cycle negative regulation of cell size positive regulation of glucose import negative regulation of Ras protein signal transduction positive regulation of insulin receptor signaling pathway response to cadmium ion intracellular distribution of mitochondria camera-type eye morphogenesis negative regulation of smooth muscle cell proliferation response to axon injury mitochondrial calcium ion homeostasis response to calcium ion response to electrical stimulus mitochondrion localization negative regulation of glial cell proliferation negative regulation of sarcomere organization positive regulation of dendritic spine morphogenesis parkin-mediated mitophagy in response to mitochondrial depolarization response to fatty acid cellular response to manganese ion cellular response to glucose stimulus cellular response to testosterone stimulus cellular response to low-density lipoprotein particle stimulus cellular response to hypoxia cellular response to ionizing radiation cellular response to dexamethasone stimulus cellular response to mercaptoethanol negative regulation of release of cytochrome c from mitochondria negative regulation of mitochondrial fission positive regulation of cold-induced thermogenesis positive regulation of cellular respiration negative regulation of reactive oxygen species biosynthetic process positive regulation of reactive oxygen species biosynthetic process response to L-arginine negative regulation of vascular smooth muscle cell proliferation positive regulation of vascular smooth muscle cell proliferation cellular response to L-glutamate response to D-galactose response to flavonoid positive regulation of vascular associated smooth muscle cell apoptotic process response to hypobaric hypoxia positive regulation of ovarian follicle development positive regulation of estradiol secretion positive regulation of progesterone secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch