Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MGEPSREEYK IQSFDAETQQ LLKTALKVAC FETEDGEYSV CQRSYSNCSR LMPSRCNTQY RDPGAVDLEK VANVIVDHSL QDCVFSKEAG RMCYAIIQAE SKQAGQSVFR RGLLNRLQQE YQAREQLRAR SLQGWVCYVT FICNIFDYLR VNNMPMMALV NPVYDCLFRL AQPDSLSKEE EVDCLVLQLH RVGEQLEKMN GQRMDELFVL IRDGFLLPTG LSSLAQLLLL EIIEFRAAGW KTTPAAHKYY YSEVSD
MIF4GD is a protein which contains an MIF4G domain.This gene encodes a protein which contains an MIF4G domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: SLBP (stem loop binding protein)-interacting protein 1; SLBP-interacting protein 1; unnamed protein product
Gene Aliases: AD023; MIFD; SLIP1
UniProt ID: (Human) A9UHW6
Entrez Gene ID: (Human) 57409
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support