    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MHC Class I (H-2Kk) Antibodies
          Invitrogen
          H-2Kk Monoclonal Antibody (36-7-5), PE
          View all (8) MHC Class I (H-2Kk) antibodies
          Questions & Answers
          Cite H-2Kk Monoclonal Antibody (36-7-5), PE
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          H-2Kk Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          H-2Kk Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          H-2Kk Antibody (MM4204) in Flow
          Flow cytometry analysis of H-2Kk in AKR mouse splenocytes. Cells were stained using H-2Kk Monoclonal Antibody (36-7-5), PE (Product # MM4204). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          H-2Kk Antibody in Flow Cytometry (Flow)
          H-2Kk Monoclonal Antibody (36-7-5), PE
          Product Details
          MM4204
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          0.2 µg/1x10^6 cells
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          36-7-5
          Immunogen
          Mouse H-2Dk. from A.AL mouse splenocytes
          3D Epitope / Immunogen
          Conjugate
          PE PE PE
          • FITC
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.1 mg/mL
          Amount
          100 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.5
          Contains
          <0.1% sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2539789
          Target Information
          This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8.
          HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: class I major histocompatibility antigen H-2Kd; H-2 class I histocompatibility antigen, K-B alpha chain; H-2 class I histocompatibility antigen, K-D alpha chain; H-2 class I histocompatibility antigen, K-K alpha chain; H-2 class I histocompatibility antigen, K-Q alpha chain; H-2 class I histocompatibility antigen, K-W28 alpha chain; H-2K(B); H-2K(K); H-2K(Q); H2DD; H2K; H2K1; H2KD; H2KK; MHC class I antigen; MHC class I H2-K-b-alpha-2 cell surface glycoprotein; MHC class I heavy chain H2-K; MHC H2-K transplantation antigen
          View more View less
          Gene Aliases: H-2K; H-2K(d); H2-K; H2-K1; K-f
          View more View less
          UniProt ID: (Mouse) P01901
          View more View less
          Entrez Gene ID: (Mouse) 14972
          View more View less
          Function(s)
          receptor binding protein binding beta-2-microglobulin binding peptide binding peptide antigen binding T cell receptor binding poly(A) RNA binding TAP binding protein heterodimerization activity major histocompatibility complex protein
          Process(es)
          positive regulation of T cell mediated cytotoxicity immune system process antigen processing and presentation of peptide antigen via MHC class I antigen processing and presentation of endogenous peptide antigen via MHC class I via ER pathway, TAP-dependent immune response antigen processing and presentation antigen processing and presentation of exogenous peptide antigen via MHC class I defense response to bacterium inner ear development
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline