    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Tau Antibodies
          Invitrogen
          Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8)
          Advanced Verification
          93 Published Figures
          1,288 References
          View all (227) Tau antibodies
          Questions & Answers
          Cite Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8)
          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:
          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (93)
          • Advanced Verification (1)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 96
          Phospho-Tau (Ser202, Thr205) Antibody (MN1020) in ICC/IF
          Immunofluorescence analysis of Phospho-Tau (Ser202, Thr205) was performed using 70% confluent log phase SH-SY5Y Retinoic acid 100 nM, 8 days. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8) (Product ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunohistochemistry, Immunocytochemistry (WB, IHC, ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry, ELISA (IHC, ELISA)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry (IHC)
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          Phospho-Tau (Ser202, Thr205) Antibody in Immunohistochemistry - Free Floating (IHC (Free))
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody in Western Blot (WB)
          Phospho-Tau (Ser202, Thr205) Antibody
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Phospho-Tau (Ser202, Thr205) Monoclonal Antibody (AT8)
          Product Details
          MN1020
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:250-1:2,000
          View 411 publications 411 publications
          Immunohistochemistry (IHC)
          -
          View 643 publications 643 publications
          Immunohistochemistry (Paraffin) (IHC (P))
          5-10 µg/mL
          View 61 publications 61 publications
          Immunohistochemistry (Frozen) (IHC (F))
          -
          View 17 publications 17 publications
          Immunohistochemistry - Free Floating (IHC (Free))
          -
          View 18 publications 18 publications
          Immunocytochemistry (ICC/IF)
          1:50-1:1,000
          View 87 publications 87 publications
          Flow Cytometry (Flow)
          -
          View 5 publications 5 publications
          ELISA (ELISA)
          -
          View 12 publications 12 publications
          Immunoprecipitation (IP)
          -
          View 2 publications 2 publications
          Proximity Ligation Assay (PLA) (PLA)
          -
          View 1 publication 1 publication
          Dot blot (DB)
          -
          View 2 publications 2 publications
          Immunomicroscopy (IM)
          -
          View 2 publications 2 publications
          Inhibition Assays (Inhibition)
          -
          View 1 publication 1 publication
          Immunodiffusion (ID)
          -
          View 1 publication 1 publication
          In vitro Assay (In vitro)
          -
          View 1 publication 1 publication
          Miscellaneous PubMed (Misc)
          -
          View 24 publications 24 publications
          Product Specifications
          Species Reactivity
          Human, Mouse
          Published species
          C. elegans, Cat, Chimpanzee, Cynomolgus monkey, Dog, Fruit fly, Hamster, Human, Monkey, Mouse, Non-human primate, Pig, Rabbit, Rat, Rhesus monkey, Yeast, Zebrafish
          Host/Isotype
          Mouse / IgG1, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          AT8
          Immunogen
          Partially purified human PHF-Tau
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          0.2 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          PBS
          Contains
          no preservative
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_223647
          Product Specific Information
          MN1020 recognizes a phosphatase-senstive epitope on PHF-Tau. No cross reactivity with normal Tau has been observed. The epitope of this antibody contains the phosphorylated Ser202 (4,5) residue (numbering according to human Tau40).
          MN1020 detects PHF-tau (Ser202/Thr205)a which has a predicted molecular weight of approximately 79 kDa.
          This product is a Low Endotoxin formulation.
          Purity is >95% as determined by SDS-PAGE.
          Target Information
          Tau is a neuronal microtubule-associated protein found predominantly on axons. The function of Tau is to promote tubulin polymerization and stabilize microtubules. The C-terminus binds axonal microtubules while the N- terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by TAU/MAPT localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton while the longer isoforms may preferentially play a role in its stabilization. In its hyper-phosphorylated form, Tau is the major component of paired helical filaments (PHF), the building block of neurofibrillary lesions in Alzheimer's diseases (AD) brain. Hyper-phosphorylation impairs the microtubule binding function of Tau, resulting in the destabilization of microtubules in AD brains, ultimately leading to the degeneration of the affected neurons. Numerous serine/threonine kinases phosphorylate Tau, including GSK-3beta, protein kinase A (PKA), cyclin-dependent kinase 5 (cdk5) and casein kinase II. Hyper-phosphorylated Tau is found in neurofibrillary lesions in a range and other central nervous system disorders such as Pick's disease, frontotemporal dementia, cortico-basal degeneration and progressive supranuclear palsy.
          Bioinformatics
          Protein Aliases: FLJ31424; FTDP17; G protein beta1/gamma2 subunit-interacting factor 1; map tau; MGC138549; microtubule-associated protein tau; microtubules; neurofibrillary tangle protein; neurofibrillary tangles; Neuronal Marker; paired helical filament-tau; PHF-tau; PHFtau; protein phosphatase 1, regulatory subunit 103; tau microtubule binding protein; Tau-4; Tau-derived paired helical filament hexapeptide; Tau5; unnamed protein product
          View more View less
          Gene Aliases: DDPAC; FTD1; FTDP-17; MAPTL; MSTD; Mtapt; MTBT1; MTBT2; PHF-tau; PPND; PPP1R103; TAU; tau-40; Tau-PHF6
          View more View less
          UniProt ID: (Human) P10636, (Mouse) P10637
          View more View less
          Entrez Gene ID: (Human) 4137, (Mouse) 17762
          View more View less
          Function(s)
          DNA binding AT DNA binding double-stranded DNA binding single-stranded DNA binding RNA binding actin binding enzyme inhibitor activity protein binding microtubule binding tubulin binding SH3 domain binding enzyme binding protein kinase binding protein binding, bridging apolipoprotein binding dynactin binding phosphatidylinositol binding identical protein binding sequence-specific DNA binding chaperone binding protein phosphatase 2A binding Hsp90 protein binding lipoprotein particle binding histone-dependent DNA binding microtubule lateral binding microtubule stabilizing activity phosphatidylinositol bisphosphate binding heat shock protein binding macromolecular complex binding
          Process(es)
          microtubule cytoskeleton organization microglial cell activation cellular response to DNA damage stimulus cell-cell signaling synapse assembly learning or memory memory response to lead ion regulation of autophagy negative regulation of gene expression negative regulation of mitochondrial membrane potential rRNA metabolic process axonal transport of mitochondrion central nervous system neuron development regulation of microtubule polymerization or depolymerization regulation of microtubule polymerization positive regulation of microtubule polymerization cytoplasmic microtubule organization neuron projection development positive regulation of superoxide anion generation regulation of chromosome organization stress granule assembly cellular response to heat cellular response to reactive oxygen species positive regulation of axon extension microtubule polymerization astrocyte activation regulation of synaptic plasticity intracellular distribution of mitochondria generation of neurons synapse organization regulation of calcium-mediated signaling protein polymerization protein homooligomerization regulation of microtubule-based movement axon development regulation of microtubule cytoskeleton organization plus-end-directed organelle transport along microtubule regulation of mitochondrial fission negative regulation of mitochondrial fission fibril organization axonal transport regulation of cellular response to heat regulation of long term synaptic depression positive regulation of protein localization to synapse neurofibrillary tangle assembly negative regulation of establishment of protein localization to mitochondrion positive regulation of cellular protein localization amyloid fibril formation cellular response to nerve growth factor stimulus cellular response to brain-derived neurotrophic factor stimulus neuron migration axonogenesis response to nutrient adult walking behavior axo-dendritic transport intrinsic apoptotic signaling pathway in response to oxidative stress positive regulation of mitochondrial membrane potential positive regulation of neuron projection development negative regulation of intracellular transport mitochondrion transport along microtubule axon extension mRNA transport response to hyperoxia positive regulation of long term synaptic depression negative regulation of tubulin deacetylation positive regulation of mesenchymal stem cell migration
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-7bf6cff7cf-9d2s2:80/100.66.76.166:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0