PA5-29175 targets Mox1 in WB applications and shows reactivity with Human samples.
The PA5-29175 immunogen is recombinant fragment corresponding to a region within amino acids 201 and 456 of Human Mox1.
MEOX1 belongs to a family of nonclustered, diverged homeobox genes. It may play a role in regulating growth and differentiation.