PA1-32854 detects GPCR MRGX4 from human samples.
PA1-32854 has been successfully used in immunohistochemistry (paraffin tissue) applications.
The PA1-32854 immunogen is: Synthetic peptide, KLH conjugated.
Orphan receptor. Probably involved in the function of nociceptive neurons. May regulate nociceptor function and/or development, including the sensation or modulation of pain. Potently activated by enkephalins.