Search
Search
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: METDAPQPGL ASPDSPHDPC KMFIGGLSWQ TTQEGLREYF GQFGEVKECL VMRDPLTKRS RGFGFVTFMD QAGVDKVLAQ SRHELDSKTI DPKVAFPRRA QPKMVTRTKK IFVGGLSVNT TVEDVKQYFE QFGKVDDAML MFDKTTNRHR GFGFVTFESE DIVEKVCEIH FHEINNKMVE CKKAQPKEVM SPTGSARGRS RVMPYGMDAF MLGIGMLGYP GFQATTYASR SYTGLAPGYT YQFPEFRVER TPLPSAPVLP ELTAIPLTAY GPMAAAAAAA AVVRGTGSHP WTMAPPPGST PSRTGGFLGT TSPGPMAELY GAANQDSGVS SYISAASPAP STGFGHSLGG PLIATAFTNG YH
Musashi-1 belongs to the Musashi family of RNA-binding proteins which play a role in cell fate determination, and maintenance of stem-cell state. Musashi family proteins are also involved in post transcriptional gene regulation. The gene encodes a protein with two conserved tandem RNA recognition motifs with highly conserved RNP (ribonucleoprotein) motifs. In mammals, Musashi-1 is expressed in fetal kidney, brain, liver and lung, and in adult brain and pancreas. In humans, the gene is located on chromosome 12.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Musashi-1; musashi1; RNA-binding protein Musashi homolog 1; unnamed protein product
Gene Aliases: MSI1
UniProt ID: (Human) O43347
Entrez Gene ID: (Human) 4440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support