Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • MYRIP Antibodies

          Abnova

          MYRIP Polyclonal Antibody, MaxPab™

          View all (5) MYRIP antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite MYRIP Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          MYRIP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          MYRIP Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          MYRIP Antibody (H00025924-B01P) in WB

          Western Blot analysis of MYRIP expression in transfected 293T cell line by MYRIP MaxPab polyclonal antibody. Lane 1: MYRIP transfected lysate(94.49 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          MYRIP Antibody in Western Blot (WB)

          Product Details

          H00025924-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          MYRIP (AAH92512.1, 1 a.a. approximately 859 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MGRKLDLSGL TDDETEHVLQ VVQRDFNLRK KEEERLSELK QKLDEEGSKC SILSKHQQFV EHCCMRCCSP FTFLVNTKRQ CGDCKFNVCK SCCSYQKHEK AWVCCVCQQA RLLRAQSLEW FYNNVKSRFK RFGSAKVLKN LYRKHRLESG ACFDILGGSL FESNLENEGS ISGSDSTFYR QSEGHSVMDT LAVALRVAEE AIEEAISKAE AYGDSLDKQN EASYLRDHKE ELTEELATTI LQKIIRKQKS KSEQQVEEEP GWPHPQSCST KVADEGTSAS PGGYRAPAAL WRSQSAFSIT GEEALKTPPV ETPSRQPRDQ GQHPRAESAL PSWKSVDRLD ETNLAPVLQS PDGNWVALKD GAPPPTRLLA KPKSGTFQAL EVASSVASAY DEMGSDSEED FDWSEALSKL CPRSRALPRN PQPQPTQAQS SDQGPIAASP SSALSPNPEA MCSDSETSSA GSSREVGHQA RLSWLQRKAP RNPAAEKMRL HGELDVNFNP QLASRETSDS SEPEEAPHTT DRRARRWRRA RLGSEEPSKE PSSPSAQLRD LDTHQVSDDL SETDISNEAR DPQTLTDTTE EKRRNRLYEL AMKMSEKETS SGEDQESEPK TESENQKESL SSEDNSQSVQ EELKKKFSAV SLCNISTEVL KVINATEELI AGSTGPWESP QVPPDRQKGM FPRGTDQVRL DEQLTSLEEN VYLAAGTVYG LETQLTELED AARCIHSGTD ETHLADLEDQ VATAAAQVHH AELQISDIES RISALTIAGL NIAPCVRFTR RRDQKQRTQV QTIDTSRQQR RKLPAPPVKA EKIETSSVTT IKTFNHNFIL QGSSTNRTKE RKGTTKDLME PALESAVMY

          Target Information

          MYRIP is a protein coding gene. Rab effector protein involved in melanosome transport. Serves as link between melanosome-bound RAB27A and the motor proteins MYO5A and MYO7A. May link RAB27A-containing vesicles to actin filaments. Functions as a protein kinase A-anchoring protein (AKAP). May act as a scaffolding protein that links PKA to components of the exocytosis machinery, thus facilitating exocytosis, including insulin release.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp586F1018; exophilin; exophilin 8; Exophilin-8; FLJ44025; MGC130034; MGC130035; Myosin-VIIa- and Rab-interacting protein; OTTHUMP00000209206; Rab effector MyRIP; SlaC2-c; slp homolog lacking C2 domains c; Slp homologue lacking C2 domains; synaptotagmin-like protein homologue lacking C2 domains-c; Synaptotagmin-like protein lacking C2 domains C; unnamed protein product

          View more View less

          Gene Aliases: MYRIP; SLAC2-C; SLAC2C

          View more View less

          UniProt ID: (Human) Q8NFW9

          View more View less

          Entrez Gene ID: (Human) 25924

          View more View less

          Function(s)
          actin binding protein binding zinc ion binding myosin binding small GTPase binding metal ion binding protein kinase A binding
          Process(es)
          intracellular protein transport positive regulation of insulin secretion
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline