Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 4
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Description: The polyclonal antibody reacts with human and mouse MyD88; the antibody was raised against residues 279-296 of human MyD88 and recognizes human and mouse antigens. MyD88, a myeloid differentiation primary response gene, is expressed in a variety of tissues and functions as an adapter molecule in the IL-1 signaling pathway involved in the inflammatory responses induced by cytokines and LPS. MyD88 associates with and recruits IRAK to the IL-1 receptor. Dominant negative mutants of MyD88 attenuate IL-1R-mediated NF-kappaB activation. MyD88 also functions as a regulator molecule for IL-18 receptor and human Toll receptor family. Targeted disruption of the MyD88 gene results in loss of cellular responses to IL-1 and IL-18, and MyD88-deficient mice lack responses to LPS.
Applications Reported: This polyclonal antibody has been reported for use in immunoblotting (WB).
Applications Tested: This polyclonal antibody has been reported for use in immunoblotting (1:500-1:1000 dilution) of MyD88 from Jurkat cell lysate as a positive control. A 35 kDa band can be detected. It is recommended that the reagent be carefully titrated for optimal performance in the assay of interest.
Purity: Greater than 90%, as determined by SDS-PAGE.
Aggregation: Less than 10%, as determined by HPLC.
Filtration: 0.2 µm post-manufacturing filtered.
Myeloid differentiation Marker 88 (MyD88) is an adaptor molecule for the Toll/IL-1R and is involved in the inflammatory response induced by IL-1, IL-18 and LPS. MyD88 recruits IRAK to IL-1 receptor complex in response to IL-1. This pathway further leads to activation of NF-kB. MyD88 has a wide tissue distribution.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mutant myeloid differentiation primary response 88; myeloid differentiation primary response 88; myeloid differentiation primary response gene (88); myeloid differentiation primary response protein 88; myleoid differentiation primary response protein; TLR adaptor MYD88; unnamed protein product
Gene Aliases: IMD68; MYD88D; WM1
UniProt ID: (Human) Q99836, (Mouse) P22366
Entrez Gene ID: (Human) 4615, (Mouse) 17874
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support