Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • CD56 (NCAM) Antibodies

          Proteintech

          NCAM1/CD56 Polyclonal Antibody

          47 References
          View all (231) CD56 (NCAM) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NCAM1/CD56 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (19)
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 19

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NCAM1/CD56 Antibody (14255-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded Human small cell lung carcinoma tissue slide using (Product # 14255-1-AP) NCAM1/CD56 antibody at dilution of 1:16,000 (under 10x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NCAM1/CD56 Antibody in Western Blot (WB)
          NCAM1/CD56 Antibody in Western Blot (WB)
          NCAM1/CD56 Antibody in Western Blot (WB)
          NCAM1/CD56 Antibody in Western Blot (WB)
          NCAM1/CD56 Antibody in Western Blot (WB)
          NCAM1/CD56 Antibody in Western Blot (WB)
          NCAM1/CD56 Antibody in Flow Cytometry (Flow)
          NCAM1/CD56 Polyclonal Antibody

          Product Details

          14255-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:4,000
          View 9 publications 9 publications

          Immunohistochemistry (IHC)

          -
          View 21 publications 21 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:800
          -

          Immunocytochemistry (ICC/IF)

          -
          View 15 publications 15 publications

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Not applicable (N/A)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NCAM1/CD56 Fusion Protein Ag5528 (30-352 aa encoded by BC047244)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.27 mg/mL

          Amount

          40.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: EISVGESKF FLCQVAGDAK DKDISWFSPN GEKLTPNQQR ISVVWNDDSS STLTIYNANI DDAGIYKCVV TGEDGSESEA TVNVKIFQKL MFKNAPTPQE FREGEDAVIV CDVVSSLPPT IIWKHKGRDV ILKKDVRFIV LSNNYLQIRG IKKTDEGTYR CEGRILARGE INFKDIQVIV NVPPTIQARQ NIVNATANLG QSVTLVCDAE GFPEPTMSWT KDGEQIEQEE DDEKYIFSDD SSQLTIKKVD KNDEAEYICI AENKAGEQDA TIHLKVFAKP KITYVENQTA MELEEQVTLT CEASGDPIPS ITWRTSTRNI SSEE (30-352 aa encoded by BC047244)

          Target Information

          CD56, also known as neural cell adhesion molecule (NCAM), is a highly glycosylated transmembrane glycoprotein of the immunoglobulin family. It plays a crucial role in cell adhesion, migration, axonal growth, pathfinding, and synaptic plasticity. CD56 is ubiquitously expressed in the nervous system in isoforms ranging from 120-180 kDa and is involved in homotypic adhesion of neural cells. It mediates interactions by binding extracellular matrix components such as laminin and integrins, with polysialic modification reducing CD56-mediated adhesion. In the hematopoietic system, CD56 is expressed on natural killer (NK) cells and a subset of T cells known as NKT cells. It is also found on most neuroectodermal-derived cell lines, tissues, and neoplasms, including retinoblastoma, medulloblastoma, astrocytomas, and neuroblastoma. CD56 serves as a widely used neuroendocrine marker with high sensitivity for neuroendocrine tumors and ovarian granulosa cell tumors. Diseases associated with CD56 dysfunction include rabies and blastic plasmacytoid dendritic cell neoplasms, highlighting its importance in both neural and immune system functions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 145 kda neural cell adhesion molecule; antigen recognized by monoclonal antibody 5.1H11; CD-56; CD56; E NCAM; membrane glycoprotein; N CAM1; N-CAM; N-CAM-1; neural cell adhesion molecule; Neural cell adhesion molecule 1; neural cell adhesion molecule secreted isoform; neural cell adhesion molecule, NCAM; sCD56; sNCAM; soluble CD56; soluble NCAM; unnamed protein product

          View more View less

          Gene Aliases: CD56; E-NCAM; MSK39; N-CAM; N-CAM-1; NCAM; NCAM-1; NCAM-C; NCAM1; NCAMC

          View more View less

          UniProt ID: (Human) P13591, (Mouse) P13595, (Rat) P13596

          View more View less

          Entrez Gene ID: (Human) 4684, (Pig) 100515564, (Mouse) 17967, (Rat) 24586

          View more View less

          Function(s)
          virus receptor activity protein binding heparin binding phosphatase binding LRR domain binding identical protein binding fibroblast growth factor receptor binding cytoskeletal protein binding
          Process(es)
          regulation of exocyst assembly cell adhesion cell surface receptor signaling pathway aging learning or memory response to lead ion response to fluoxetine thalamus development neuron projection development multicellular organismal response to stress homotypic cell-cell adhesion response to cocaine neuron development positive regulation of calcium-mediated signaling regulation of sensory perception of pain positive regulation of cardiac muscle cell proliferation negative regulation of cell death epithelial to mesenchymal transition viral entry into host cell commissural neuron axon guidance regulation of semaphorin-plexin signaling pathway response to inorganic substance peripheral nervous system axon regeneration calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules organ regeneration cellular response to molecule of bacterial origin cellular response to inorganic substance
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline