Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: NADEGEPGT CKDREILRHD PHKLLEGCLV GGRAMGARAA YIYIRGEFYN EASNLQVAIR EAYEAGLIGK NACGSGYDFD VFVVRGAGAY ICGEETALIE SIEGKQGKPR LKPPFPADVG VFGCPTTVAN VETVAVSPTI CRRGGTWFAG FGRERNSGTK LFNISGHVNH PCTVEEEMSV PLKELIEKHA GGVTGGWDNL LAVIPGGSST PLIPKSVCET VLMDFDALVQ AQTGLGTAAV IVMDRSTDIV KAIARLIEFY KHESCGQCTP CREGVDWMNK VMARFVRGDA RPAEIDSLWE ISKQIEGHTI CALGDGAAWP VQGLIRHFRP ELEERMQRFA QQHQARQAAS (116-464 aa encoded by BC015645)
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 51 kda flavoprotein; complex I 51 kda subunit; complex I 51kDa subunit; complex I, mitochondrial respiratory chain; complex I-51kD; mitochondrial NADH dehydrogenase ubiquinone flavoprotein 1; mitochondrial NADH:ubiquinone oxidoreductase 51 kda subunit; NADH dehydrogenase (ubiquinone) flavoprotein 1; NADH dehydrogenase (ubiquinone) flavoprotein 1, 51kDa; NADH dehydrogenase flavoprotein 1; 24 kDa (FP); NADH-ubiquinone oxidoreductase 51 kDa subunit; NDUFV1; unnamed protein product
Gene Aliases: CI-51K; CI-51kD; CI51KD; MC1DN4; UQOR1
UniProt ID: (Human) P49821, (Mouse) Q91YT0
Entrez Gene ID: (Human) 4723, (Rat) 293655, (Mouse) 17995
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support