Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NALP2 Antibodies

          Abnova

          NLRP2 Polyclonal Antibody, MaxPab™

          Greener Choice
          View all (14) NALP2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NLRP2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          NLRP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          NLRP2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NLRP2 Antibody (H00055655-B01P) in WB

          Western Blot analysis of NLRP2 expression in transfected 293T cell line by NLRP2 MaxPab polyclonal antibody. Lane 1: NLRP2 transfected lysate(116.82 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NLRP2 Antibody in Western Blot (WB)
          NLRP2 Polyclonal Antibody, MaxPab™

          Product Details

          H00055655-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NLRP2 (NP_060322.1, 1 a.a. approximately 1062 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MVSSAQMGFN LQALLEQLSQ DELSKFKYLI TTFSLAHELQ KIPHKEVDKA DGKQLVEILT THCDSYWVEM ASLQVFEKMH RMDLSERAKD EVREAALKSF NKRKPLSLGI TRKERPPLDV DEMLERFKTE AQAFTETKGN VICLGKEVFK GKKPDKDNRC RYILKTKFRE MWKSWPGDSK EVQVMAERYK MLIPFSNPRV LPGPFSYTVV LYGPAGLGKT TLAQKLMLDW AEDNLIHKFK YAFYLSCREL SRLGPCSFAE LVFRDWPELQ DDIPHILAQA RKILFVIDGF DELGAAPGAL IEDICGDWEK KKPVPVLLGS LLNRVMLPKA ALLVTTRPRA LRDLRILAEE PIYIRVEGFL EEDRRAYFLR HFGDEDQAMR AFELMRSNAA LFQLGSAPAV CWIVCTTLKL QMEKGEDPVP TCLTRTGLFL RFLCSRFPQG AQLRGALRTL SLLAAQGLWA QTSVLHREDL ERLGVQESDL RLFLDGDILR QDRVSKGCYS FIHLSFQQFL TALFYTLEKE EEEDRDGHTW DIGDVQKLLS GVERLRNPDL IQAGYYSFGL ANEKRAKELE ATFGCRMSPD IKQELLRCDI SCKGGHSTVT DLQELLGCLY ESQEEELVKE VMAQFKEISL HLNAVDVVPS SFCVKHCRNL QKMSLQVIKE NLPENVTASE SDAEVERSQD DQHMLPFWTD LCSIFGSNKD LMGLAINDSF LSASLVRILC EQIASDTCHL QRVVFKNISP ADAHRNLCLA LRGHKTVTYL TLQGNDQDDM FPALCEVLRH PECNLRYLGL VSCSATTQQW ADLSLALEVN QSLTCVNLSD NELLDEGAKL LYTTLRHPKC FLQRLSLENC HLTEANCKDL AAVLVVSREL THLCLAKNPI GNTGVKFLCE GLRYPECKLQ TLVLWNCDIT SDGCCDLTKL LQEKSSLLCL DLGLNHIGVK GMKFLCEALR KPLCNLRCLW LWGCSIPPFS CEDLCSALSC NQSLVTLDLG QNPLGSSGVK MLFETLTCSS GTLRTLRLKI DDFNDELNKL LEEIEEKNPQ LIIDTEKHHP WAERPSSHDF MI

          Target Information

          NALP2 belongs to a family of cytoplasmic proteins that have been implicated in cell responses to apoptotic and inflammatory stimuli. Unlike the prototypical NALP protein NALP1, NALP2 only contains a NACHT domain, leucine rich repeat (LRR), and pyrin-containing domain (PYD). This protein interacts with the adapter protein ASC in addition to CARD8 and caspase-1 to form an inflammasome with high proIL-1-b-processing activity and is thought to function as a modulator of NF-kappa-B and procaspase-1 activation in macrophages. It has also been suggested that NALP2, in addition to other NALP family members, can act as innate sensors of pathogens in a manner similar to the toll-like receptors (TLRs). At least two alternatively spliced transcript variants encoding distinct isoforms have been found for NALP2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: contains NACHT, LRR and PYD domains; FLJ20510; NACHT, leucine rich repeat and PYD containing 2; NACHT, LRR and PYD domains-containing protein 2; NBS1; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 2; Nucleotide-binding site protein 1; PYPAF2; PYRIN domain and NACHT domain-containing protein 1; PYRIN-containing APAF1-like protein 2; unnamed protein product

          View more View less

          Gene Aliases: CLR19.9; NALP2; NBS1; NLRP2; OZEMA18; PAN1; PYPAF2

          View more View less

          UniProt ID: (Human) Q9NX02

          View more View less

          Entrez Gene ID: (Human) 55655

          View more View less

          Function(s)
          nucleotide binding protein binding ATP binding Pyrin domain binding
          Process(es)
          immune system process apoptotic process inflammatory response regulation of interleukin-1 beta production positive regulation of interleukin-1 beta production innate immune response regulation of inflammatory response negative regulation of NIK/NF-kappaB signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline