Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Description: This MNH3-21 monoclonal antibody reacts with the extracellular domain of human Notch3, one of four members of the Notch family of receptors. Notch receptors are 300-kDa single-pass transmembrane proteins. While the extracellular domain contains numerous epidermal growth factor-like repeats for ligand binding, the intracellular domain is involved in cell signaling. Upon binding its membrane-bound ligand (either Delta or Jagged), the Notch receptor undergoes proteolytic cleavage, first by ADAM-family metalloproteases and then by gamma-secretase. The second cleavage event releases the Notch intracellular domain (NICD), which subsequently translocates into the nucleus, heterodimerizes with the DNA-binding protein RBP-J, recruits co-activator molecules, and ultimately activates transcription.
Notch3 expression has been demonstrated on some thymocyte subsets, including CD4-CD8- and CD8SP cells. This Notch receptor is also expressed on vascular smooth muscle and cells of the central nervous system. In addition to its role in stem cell hematopoiesis, Notch3 plays a pivotal role in T cell lineage commitment and thymocyte development. Moreover, Notch3 is overexpressed in human T-cell acute lymphoblastic leukemias (T-ALL) and other cancers. Finally, mutation of Notch3 has been linked to cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL), a CNS degenerative disorder.
Applications Reported: This MHN3-21 antibody has been reported for use in flow cytometric analysis.
Applications Tested: This MHN3-21 antibody has been pre-titrated and tested by flow cytometric analysis of Jurkat cells. This can be used at 5 µL (0.25 µg) per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.
Filtration: 0.2 µm post-manufacturing filtered.
NOTCH3 is the third discovered human homologue of the Drosophilia melanogaster type I membrane protein notch. In Drosophilia, notch interaction (with its cell-bound ligands delta, serrate) establishes an intercellular signalling pathway that plays a key role in neural development. Homologues of the notch-ligands have also been identified in human. NOTCH3 functions as a receptor for membrane-bound ligands Jagged1, Jagged2, and Delta1 to regulate cell-fate determination. Mutations in NOTCH3 have been identified as the underlying cause of cerebral autosomal dominant arteriopathy with subcortical infarcts and leukoencephalopathy (CADASIL).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neurogenic locus notch homolog protein 3; Notch 3; Notch homolog 3
Gene Aliases: CADASIL; CADASIL1; CASIL; IMF2; LMNS; NOTCH3
UniProt ID: (Human) Q9UM47
Entrez Gene ID: (Human) 4854
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support