Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: CVKVVQEYE RVIIFRLGHL LPGRAKGPGL FFFLPCLDTY HKVDLRLQTL EIPFHEVALD SVTCIWGIKV ERIEIKDVRL PAGLQHSLAV EAEAQRQAKV RMIAAEAEKA ASESLRMAAE ILSGTPAAVQ LRYLHTLQSL STEKPSTVVL PLPFDLLNCL SSPSNRTQGS LPFPSPSKPV EPLNPKKKDS PML (124-315 aa encoded by BC029141)
The pathological conditions of SRN1 correlate well with mutations at the NPHS2 gene, where expression of a protein known as Podocin occurs. Abnormal or inefficient signaling through Podocin protein-dependent networks contributes to the development of podocyte dysfunction and proteinuria. The human NPHS2 gene maps to chromosome 1q25-q31 and encodes a 383 amino acid protein. Podocin is an integral membrane protein that appears to fold into a hairpin-like structure with intracellular amino- and carboxy-termini. Transmembrane and cytoplasmic portions of Podocin share homology to the corresponding regions of the stomatin family proteins. Expression of high-order oligomers of Podocin in glomerular podocytes may reflect a scaffolding function that influences proper function of the glomerular filtration barrier, which is necessary for renal stability.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: nephrosis 2 homolog, podocin; nephrosis 2, idiopathic, steroid-resistant; NPHS2; NPHS2, podocin; unnamed protein product
Gene Aliases: PDCN; podocin; SRN1
UniProt ID: (Rat) Q8K4G9, (Mouse) Q91X05
Entrez Gene ID: (Rat) 170672, (Mouse) 170484
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support