Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NRG1 Antibodies

          Bioss

          NRG1 Polyclonal Antibody

          View all (32) NRG1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NRG1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          NRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          NRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NRG1 Antibody (BS-11917R) in IHC (P)

          Tissue/cell: Rat brain tissue; 4% Paraformaldehyde-fixed and paraffin-embedded; Antigen retrieval: citrate buffer (0.01 M, pH 6.0), Boiling bathing for 15 min; Blocking buffer (normal goat serum) at 37°C for 20 min; Incubation: NRG1 Polyclonal Antibody, Unconjugated (bs-11917R) 1:200, Overnight at 4°C; The secondary antibody was Goat Anti-Rabbit IgG, Cy3 conjugated (bs-0295G-Cy3) used at 1:200 dilution for ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NRG1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NRG1 Polyclonal Antibody

          Product Details

          BS-11917R

          Applications
          Tested Dilution
          Publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -

          Flow Cytometry (Flow)

          1:20-1:100
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human Neuregulin-1, amino acids 81-200.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Neuregulin 1 (NRG1) was originally identified as a 44-kD glycoprotein that interacts with the NEU/ERBB2 receptor tyrosine kinase to increase its phosphorylation on tyrosine residues. It is known that an extraordinary variety of different isoforms are produced from the NRG1 gene by alternative splicing. These isoforms include heregulins (HRGs), glial growth factors (GGFs) and sensory and motor neuron-derived factor (SMDF). They are tissue-specifically expressed and differ significantly in their structure. The HRG isoforms all contain immunoglobulin (Ig) and epidermal growth factor-like (EGF-like) domains. GGF and GGF2 isoforms contain a kringle-like sequence plus Ig and EGF-like domains; and the SMDF isoform shares only the EGF-like domain with other isoforms. The receptors for all NRG1 isoforms are the ERBB family of tyrosine kinase transmembrane receptors. Through interaction with ERBB receptors, NRG1 isoforms induce the growth and differentiation of epithelial, neuronal, glial, and other types of cells.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: acetylcholine receptor-inducing activity; glial growth factor 2; growth factor; heregulin; heregulin, alpha (45kD, ERBB2 p185-activator); neu differentiation factor; neuregulin 1; neuregulin 1 type III beta 3; NRG-1; NRG1; NRG1 class VII isoform alpha 2a; NRG1 class VII isoform alpha 2b; NRG1 class VII isoform alpha beta 2a; NRG1 class VII isoform alpha beta 3; NRG1 class VII isoform beta 2a; NRG1 class VII isoform beta 3; Pro-neuregulin-1, membrane-bound isoform; Pro-NRG1; sensory and motor neuron derived factor; type I neuregulin 1; type III neuregulin 1; unnamed protein product

          View more View less

          Gene Aliases: 6030402G23Rik; ARIA; D230005F13Rik; GGF; GGF2; GGFII; HGL; HRG; HRG1; HRGA; HRGalpha; MST131; MSTP131; NDF; NRG1; NRG1-IT2; SMDF

          View more View less

          UniProt ID: (Human) Q02297, (Mouse) Q6DR98

          View more View less

          Entrez Gene ID: (Human) 3084, (Rat) 112400, (Mouse) 211323

          View more View less

          Function(s)
          transcription cofactor activity receptor binding cytokine activity integrin binding protein binding growth factor activity zinc ion binding nickel cation binding protein tyrosine kinase activator activity transmembrane receptor protein tyrosine kinase activator activity receptor tyrosine kinase binding ErbB-3 class receptor binding chemorepellent activity receptor agonist activity ErbB-2 class receptor binding growth factor
          Process(es)
          ventricular trabecula myocardium morphogenesis cell communication transmembrane receptor protein tyrosine kinase signaling pathway nervous system development brain development peripheral nervous system development cell proliferation positive regulation of cell proliferation regulation of cell communication negative regulation of cardiac muscle cell apoptotic process neural crest cell development regulation of signaling cell differentiation positive regulation of cell growth mammary gland development positive regulation of protein complex assembly activation of protein kinase B activity intracellular signal transduction ERBB signaling pathway ERBB2 signaling pathway ERBB3 signaling pathway ERBB4 signaling pathway ERBB2-ERBB3 signaling pathway ERBB2-ERBB4 signaling pathway ERBB4-ERBB4 signaling pathway wound healing positive regulation of cell adhesion negative regulation of transcription, DNA-templated negative chemotaxis negative regulation of secretion positive regulation of developmental process positive regulation of striated muscle cell differentiation cardiac muscle cell differentiation ventricular cardiac muscle cell differentiation positive regulation of cardiac muscle cell proliferation cardiac muscle cell myoblast differentiation endocardial cell differentiation regulation of biological quality positive regulation of ERK1 and ERK2 cascade regulation of postsynaptic neurotransmitter receptor internalization positive regulation of peptidyl-tyrosine autophosphorylation negative regulation of extrinsic apoptotic signaling pathway in absence of ligand MAPK cascade cell morphogenesis cardiac conduction system development activation of transmembrane receptor protein tyrosine kinase activity synapse assembly heart development muscle organ development locomotory behavior regulation of blood pressure glial cell differentiation glucose transport cell migration glial cell fate commitment negative regulation of protein catabolic process myelination regulation of protein heterodimerization activity cellular protein complex disassembly neurotransmitter receptor metabolic process regulation of cell differentiation positive regulation of protein kinase activity positive regulation of Ras protein signal transduction animal organ development neuron fate commitment positive regulation of protein kinase B signaling positive regulation of protein tyrosine kinase activity positive regulation of calcineurin-NFAT signaling cascade positive regulation of cardiac muscle cell differentiation axon ensheathment positive regulation of myelination oligodendrocyte differentiation
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline