Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NSDHL Antibodies

          Proteintech

          NSDHL Polyclonal Antibody

          14 References
          View all (13) NSDHL antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite NSDHL Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (24)
          NSDHL Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NSDHL Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 24

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NSDHL Antibody (15111-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol ) fixed HeLa cells using 15111-1-AP (NSDHL antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NSDHL Antibody in Immunocytochemistry (ICC/IF)
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NSDHL Antibody in Western Blot (WB)
          NSDHL Antibody in Western Blot (WB)
          NSDHL Antibody in Western Blot (WB)
          NSDHL Polyclonal Antibody

          Product Details

          15111-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,200
          View 8 publications 8 publications

          Immunohistochemistry (IHC)

          -
          View 6 publications 6 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          NSDHL Fusion Protein Ag7097 (1-303 aa encoded by BC000245)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.39 mg/mL

          Amount

          58.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MEPAVSEPM RDQVARTHLT EDTPKVNADI EKVNQNQAKR CTVIGGSGFL GQHMVEQLLA RGYAVNVFDI QQGFDNPQVR FFLGDLCSRQ DLYPALKGVN TVFHCASPPP SSNNKELFYR VNYIGTKNVI ETCKEAGVQK LILTSSASVI FEGVDIKNGT EDLPYAMKPI DYYTETKILQ ERAVLGANDP EKNFLTTAIR PHGIFGPRDP QLVPILIEAA RNGKMKFVIG NGKNLVDFTF VENVVHGHIL AAEQLSRDST LGGKAFHITN DEPIPFWTFL SRILTGLNYE APKYHIPYWV AYYL (1-303 aa encoded by BC000245)

          Target Information

          The protein encoded by this gene is localized in the endoplasmic reticulum and is involved in cholesterol biosynthesis. Mutations in this gene are associated with CHILD syndrome, which is a X-linked dominant disorder of lipid metabolism with disturbed cholesterol biosynthesis, and typically lethal in males. Alternatively spliced transcript variants with differing 5' UTR have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 3-beta-hydroxysteroid dehydrogenase; 3beta-hydroxysteroid-4alpha-carboxylate 3-dehydrogenase (decarboxylating); bare patches; epididymis secretory sperm binding protein; H105e3; NAD(P) dependent steroid dehydrogenase-like protein transcript; NAD(P)-dependent steroid dehydrogenase-like protein; Protein H105e3; short chain dehydrogenase/reductase family 31E, member 1; Sterol-4-alpha-carboxylate 3-dehydrogenase, decarboxylating; striated

          View more View less

          Gene Aliases: AI747449; Bpa; H105E3; NSDHL; SDR31E1; Str; XAP104

          View more View less

          UniProt ID: (Human) Q15738, (Mouse) Q9R1J0, (Rat) Q5PPL3

          View more View less

          Entrez Gene ID: (Human) 50814, (Mouse) 18194, (Rat) 309262

          View more View less

          Function(s)
          C-3 sterol dehydrogenase (C-4 sterol decarboxylase) activity 3-beta-hydroxy-delta5-steroid dehydrogenase activity protein binding oxidoreductase activity oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor 3-beta-hydroxysteroid dehydrogenase (NAD+)/C4-decarboxylase activity sterol-4-alpha-carboxylate 3-dehydrogenase (decarboxylating) activity molecular_function dehydratase
          Process(es)
          hair follicle development lipid metabolic process steroid biosynthetic process cholesterol biosynthetic process smoothened signaling pathway steroid metabolic process cholesterol metabolic process sterol biosynthetic process labyrinthine layer blood vessel development oxidation-reduction process biological_process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline