Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
A suggested positive control for this product is MDCKII cell lysate.
Netrin-1 is a protein target that plays a crucial role in various biological processes, particularly in the development and guidance of neuronal cells. It belongs to the netrin family of axon guidance molecules and is involved in mediating axon guidance, cell migration, and tissue morphogenesis during embryonic development. Netrin-1 functions by binding to specific receptors on the cell surface and providing directional cues for cellular movement and connectivity. Dysregulation of Netrin-1 has been implicated in various diseases, including cancer, neurological disorders, and cardiovascular conditions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis tissue protein Li 131P; HGNC:8029; netrin 1, mouse, homolog of; Netrin1; tropic factor
Gene Aliases: MRMV4; NET1; Netrin-1; NTN1L
UniProt ID: (Human) O95631, (Mouse) O09118
Entrez Gene ID: (Human) 9423, (Bovine) 522767, (Mouse) 18208, (Rat) 114523
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support