Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • NF1 Antibodies

          OriGene

          Neurofibromin 1 Polyclonal Antibody

          View all (23) NF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Neurofibromin 1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Neurofibromin 1 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          Neurofibromin 1 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Neurofibromin 1 Antibody (TA590747) in ELISA

          ELISA: Neurofibromin 1 Antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Neurofibromin 1 Antibody in ELISA (ELISA)
          Neurofibromin 1 Polyclonal Antibody

          Product Details

          TA590747

          Applications
          Tested Dilution
          Publications

          ELISA (ELISA)

          1:100-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          DNA immunization. This antibody is specific for the N Terminus Region of the target protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.26 mg/mL

          Amount

          100 µg

          Purification

          Protein A/G

          Storage buffer

          PBS

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The protein encoded by this gene is a type I integral membrane protein localized in the intermediate region between the endoplasmic reticulum and the Golgi, presumably recycling between the two compartments. The protein is a mannose-specific lectin and is a member of a novel family of plant lectin homologs in the secretory pathway of animal cells. Mutations in the gene are associated with a coagulation defect. Using positional cloning, the gene was identified as the disease gene leading to combined factor V-factor VIII deficiency, a rare, autosomal recessive disorder in which both coagulation factors V and VIII are diminished.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp686J1293; FLJ21220; neurofibromatosis 1; Neurofibromatosis-related protein NF-1; Neurofibromin; unnamed protein product

          View more View less

          Gene Aliases: NF1; NFNS; VRNF; WSS

          View more View less

          UniProt ID: (Human) P21359

          View more View less

          Entrez Gene ID: (Human) 4763

          View more View less

          Function(s)
          GTPase activator activity protein binding lipid binding phosphatidylethanolamine binding phosphatidylcholine binding
          Process(es)
          MAPK cascade angiogenesis osteoblast differentiation metanephros development response to hypoxia liver development endothelial cell proliferation negative regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation regulation of cell-matrix adhesion negative regulation of cell-matrix adhesion negative regulation of leukocyte migration protein import into nucleus apoptotic process cell communication Ras protein signal transduction neurotransmitter secretion neuroblast proliferation negative regulation of neuroblast proliferation brain development peripheral nervous system development heart development skeletal muscle tissue development cell proliferation negative regulation of cell proliferation visual learning extrinsic apoptotic signaling pathway via death domain receptors regulation of gene expression negative regulation of Schwann cell proliferation glial cell proliferation Schwann cell proliferation Schwann cell development cell migration negative regulation of angiogenesis Rac protein signal transduction spinal cord development amygdala development forebrain astrocyte development neural tube development cerebral cortex development myelination in peripheral nervous system actin cytoskeleton organization extracellular matrix organization collagen fibril organization osteoclast differentiation adrenal gland development negative regulation of cell migration regulation of synaptic transmission, GABAergic mast cell apoptotic process positive regulation of mast cell apoptotic process myeloid cell apoptotic process positive regulation of myeloid cell apoptotic process cellular response to heat negative regulation of Rac protein signal transduction Schwann cell migration wound healing regulation of cell proliferation negative regulation of protein import into nucleus positive regulation of apoptotic process regulation of MAPK cascade negative regulation of MAPK cascade pigmentation protein kinase B signaling positive regulation of neuron apoptotic process regulation of blood vessel endothelial cell migration positive regulation of GTPase activity regulation of bone resorption negative regulation of osteoclast differentiation regulation of glial cell differentiation regulation of angiogenesis negative regulation of Ras protein signal transduction negative regulation of neurotransmitter secretion fibroblast proliferation negative regulation of fibroblast proliferation regulation of long-term neuronal synaptic plasticity sympathetic nervous system development camera-type eye morphogenesis astrocyte differentiation oligodendrocyte differentiation negative regulation of astrocyte differentiation negative regulation of oligodendrocyte differentiation smooth muscle tissue development hair follicle maturation artery morphogenesis forebrain morphogenesis regulation of developmental process cognition neuron apoptotic process negative regulation of glial cell proliferation long-term synaptic potentiation gamma-aminobutyric acid secretion, neurotransmission glutamate secretion, neurotransmission regulation of ERK1 and ERK2 cascade mast cell proliferation negative regulation of mast cell proliferation leukocyte apoptotic process stem cell proliferation extrinsic apoptotic signaling pathway in absence of ligand myeloid leukocyte migration observational learning regulation of postsynapse organization negative regulation of Schwann cell migration regulation of long-term synaptic potentiation regulation of intracellular signal transduction positive regulation of vascular smooth muscle cell proliferation vascular associated smooth muscle cell migration negative regulation of vascular associated smooth muscle cell migration vascular associated smooth muscle cell proliferation positive regulation of leukocyte apoptotic process negative regulation of stem cell proliferation positive regulation of extrinsic apoptotic signaling pathway in absence of ligand
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0