Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • TACR1 Antibodies

          Bioss

          Neurokinin 1 Receptor Polyclonal Antibody

          View all (26) TACR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Neurokinin 1 Receptor Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (4)
          Neurokinin 1 Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Neurokinin 1 Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Neurokinin 1 Receptor Antibody (BS-0064R) in IHC (P)

          Formalin-fixed and paraffin embedded rat brain labeled with Anti Substance P Receptor/NK1R Polyclonal Antibody, Unconjugated (bs-0064R) at 1:200 followed by conjugation to the secondary antibody Goat Anti-Rabbit IgG, PE conjugated (bs-0295G-PE)used at 1:200 dilution for 40 minutes at 37°C. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Neurokinin 1 Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neurokinin 1 Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Neurokinin 1 Receptor Antibody in Western Blot (WB)
          Neurokinin 1 Receptor Antibody in Western Blot (WB)
          Neurokinin 1 Receptor Polyclonal Antibody

          Product Details

          BS-0064R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human Neurokinin 1 Receptor, amino acids 2-50.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          Tachykinin Receptor 1, a member of the Tachykinin Receptor subfamily, is involved in the mediation of phosphatidylinositol metabolism by the tachykinin substance P, also referred to as neurokinin 1. It is associated with the management of depression, anxiety, and stress. It is also involved in regulating behavioral responses, neuronal survival, and cardiovascular and respiratory functions. The tachykinin receptor 1 is implicated in the progression of rheumatoid arthritis and osteoarthritis. Two variants of the tachykinin receptor 1 are produced by alternative splicing. Tachykinin receptor 1 has been reported primarily in brain, and in blood lymphocytes, nose, small intestine, and stomach. ESTs have been isolated from brain and spleen libraries.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: G-protein coupled receptor; substance P receptor; neurokinin 1 receptor; neurokinin receptor 1; NK-1 receptor; NK-1R; Nk1 receptor; SPR; substance P receptor; Substance-P receptor; Tachykinin receptor 1; tachykinin receptor 1 (substance P receptor; neurokinin-1 receptor); unnamed protein product

          View more View less

          Gene Aliases: NK1R; NKIR; SPR; TAC1R; TACR1

          View more View less

          UniProt ID: (Human) P25103, (Mouse) P30548, (Rat) P14600

          View more View less

          Entrez Gene ID: (Human) 6869, (Mouse) 21336, (Rat) 24807

          View more View less

          Function(s)
          G-protein coupled receptor activity tachykinin receptor activity protein binding substance P receptor activity signal transducer activity G-protein coupled receptor
          Process(es)
          aggressive behavior positive regulation of leukocyte migration angiotensin-mediated drinking behavior inflammatory response signal transduction G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway positive regulation of cytosolic calcium ion concentration tachykinin receptor signaling pathway learning or memory long-term memory regulation of blood pressure associative learning detection of abiotic stimulus response to hormone response to ozone positive regulation of epithelial cell migration response to auditory stimulus regulation of smooth muscle cell migration positive regulation of synaptic transmission, cholinergic positive regulation of synaptic transmission, GABAergic response to estradiol response to progesterone response to nicotine operant conditioning sperm ejaculation eating behavior positive regulation of vascular permeability response to ethanol positive regulation of action potential positive regulation of blood pressure positive regulation of ossification positive regulation of vasoconstriction positive regulation of hormone secretion response to pain behavioral response to pain regulation of smooth muscle cell proliferation positive regulation of lymphocyte proliferation positive regulation of epithelial cell proliferation positive regulation of stress fiber assembly response to electrical stimulus smooth muscle contraction involved in micturition regulation of uterine smooth muscle contraction positive regulation of uterine smooth muscle contraction positive regulation of sperm motility neuropeptide signaling pathway chemical synaptic transmission response to heat response to organic cyclic compound sensory perception of pain positive regulation of renal sodium excretion positive regulation of saliva secretion acute inflammatory response response to morphine
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0