Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Nurr1 Antibodies

          Bioss

          Nurr1 Polyclonal Antibody

          View all (36) Nurr1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Nurr1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          Nurr1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Nurr1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Nurr1 Antibody (BS-20744R) in WB

          Mouse Adrenal Gland lysate at 40µg; Probed with Nurr1 Polyclonal Antibody, Unconjugated (bs-20744R) at 1:300 overnight at 4°C followed by incubation with a conjugated secondary antibody at 1:20000 for 60 minutes at 37°C. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Nurr1 Antibody in Western Blot (WB)
          Nurr1 Polyclonal Antibody

          Product Details

          BS-20744R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200-1:400
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from human Nurr1, amino acids 331-430.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The Nuclear receptor related 1 protein (Nurr1) also known as NR4A2 (nuclear receptor subfamily 4, group A, member 2) is a protein that in humans is encoded by the NR4A2 gene. NURR1 is a member of the nuclear receptor family of intracellular transcription factors.Nurr1 plays a key role in the maintenance of the dopaminergic system of the brain. Mutations in this gene have been associated with disorders related to dopaminergic dysfunction, including Parkinson's disease, schizophrenia, and manic depression. Misregulation of this gene may be associated with rheumatoid arthritis. Four transcript variants encoding four distinct isoforms have been identified for this gene. Additional alternate splice variants may exist, but their full-length nature has not been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HZF3; Immediate-early response protein NOT; intermediate-early receptor protein; NGFI-B; NGFI-B beta; NGFI-B/nur77 beta-type transcription factor homolog; NOT; Nuclear orphan receptor HZF-3; nuclear receptor related 1; Nuclear receptor subfamily 4 group A member 2; nuclear receptor subfamily 4 group A member 2 variant TINUR; nuclear receptor subfamily 4 group A member 2 variant NURR1a; nuclear receptor subfamily 4 group A member 2 variant NURR1b; nuclear receptor subfamily 4 group A member 2 variant NURR1c; nuclear receptor subfamily 4 group A member 2 variant NURR2c; nur related protein 1; nur related protein-1, human homolog of; NUR-related factor 1; orphan nuclear receptor NR4A2; Orphan nuclear receptor NURR1; putative new variant; Regenerating liver nuclear receptor 1; RNR-1; SL-322; T-cell nuclear receptor NOT; transcriptionally inducible nuclear receptor related 1; Transcriptionally-inducible nuclear receptor; unnamed protein product

          View more View less

          Gene Aliases: HZF-3; IDLDP; NOT; NR4A2; NURR1; RNR-1; RNR1; TINOR; TINUR

          View more View less

          UniProt ID: (Human) P43354, (Mouse) Q06219, (Rat) Q07917

          View more View less

          Entrez Gene ID: (Human) 4929, (Mouse) 18227, (Rat) 54278

          View more View less

          Function(s)
          RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding DNA binding transcription factor activity, sequence-specific DNA binding RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding protein binding beta-catenin binding zinc ion binding glucocorticoid receptor binding sequence-specific DNA binding metal ion binding retinoid X receptor binding protein heterodimerization activity sequence-specific double-stranded DNA binding RNA polymerase II core promoter sequence-specific DNA binding transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding steroid hormone receptor activity C4 zinc finger nuclear receptor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter response to hypoxia neuron migration response to amphetamine transcription, DNA-templated regulation of transcription, DNA-templated regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter nervous system development adult locomotory behavior post-embryonic development gene expression regulation of gene expression central nervous system projection neuron axonogenesis central nervous system neuron differentiation habenula development intracellular receptor signaling pathway cellular response to oxidative stress regulation of dopamine metabolic process dopamine biosynthetic process dopamine metabolic process neuron maturation negative regulation of neuron apoptotic process regulation of respiratory gaseous exchange fat cell differentiation positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter neuron apoptotic process general adaptation syndrome canonical Wnt signaling pathway cellular response to corticotropin-releasing hormone stimulus dopaminergic neuron differentiation midbrain dopaminergic neuron differentiation negative regulation of apoptotic signaling pathway response to inorganic substance obsolete death response to insecticide neuron differentiation cellular response to extracellular stimulus positive regulation of catalytic activity steroid hormone mediated signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline