Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • OAS1 Antibodies

          Proteintech

          OAS1 Polyclonal Antibody

          25 References
          View all (9) OAS1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite OAS1 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (7)
          OAS1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          OAS1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OAS1 Antibody (14955-1-AP) in ICC/IF

          Immunofluorescent analysis of ( 10% Formaldehyde ) fixed HeLa cells using 14955-1-AP (OAS1 antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OAS1 Antibody in Immunocytochemistry (ICC/IF)
          OAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          OAS1 Antibody in Western Blot (WB)
          OAS1 Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          OAS1 Polyclonal Antibody

          Product Details

          14955-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 18 publications 18 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:100-1:400
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Pig

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          OAS1 Fusion Protein Ag6793 (1-351 aa encoded by BC000562)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.16 mg/mL

          Amount

          24 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MMDLRNTPA KSLDKFIEDY LLPDTCFRMQ INHAIDIICG FLKERCFRGS SYPVCVSKVV KGGSSGKGTT LRGRSDADLV VFLSPLTTFQ DQLNRRGEFI QEIRRQLEAC QRERAFSVKF EVQAPRWGNP RALSFVLSSL QLGEGVEFDV LPAFDALGQL TGSYKPNPQI YVKLIEECTD LQKEGEFSTC FTELQRDFLK QRPTKLKSLI RLVKHWYQNC KKKLGKLPPQ YALELLTVYA WERGSMKTHF NTAQGFRTVL ELVINYQQLC IYWTKYYDFK NPIIEKYLRR QLTKPRPVIL DPADPTGNLG GGDPKGWRQL AQEAEAWLNY PCFKNWDGSP VSSWILLVRP PA (1-351 aa encoded by BC000562)

          Target Information

          OAS1 is an interferon inducible protein that may play a role in mediating resistance to virus infection, control of cell growth, differentiation, and apoptosis. It binds double-stranded RNA and polymerizes ATP into PPP(A2'P5'A)N oligomers, which activate the latent RNase L that, when activated, cleaves single-stranded RNAs. This protein is associated with different subcellular fractions such as mitochondrial, nuclear, and rough/smooth microsomal fractions.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: (2-5')oligo(A) synthase 1; (2-5')oligo(A) synthase 1A; (2-5')oligo(A) synthetase 1; (2-5')oligo(A) synthetase 1A; 2',5'-oligo A synthetase 1; 2',5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5' oligo A synthetase; 2'-5' oligoadenylate synthetase 1G; 2'-5'-oligoadenylate synthase 1; 2'-5'-oligoadenylate synthetase 1; 2'-5'-oligoadenylate synthetase 1, 40/46kDa; 2'-5'-oligoisoadenylate synthetase 1; 2-5A synthase 1; 2-5A synthase 1A; 2-5A synthetase 1; 2-5A synthetase 1A; 25 oligoadenylate synthetase; E16 (2'-5') oligo A synthetase; IFI 4; IFI4; OAS 1; p42 OAS; short exon 3; targeted by nonsense-mediated decay (NMD) due to premature termination codon in exon 4; synthetase; unnamed protein product

          View more View less

          Gene Aliases: E18/E16; IFI-4; IMD100; L3; Oas1; Oas1g; OIAS; OIASI

          View more View less

          UniProt ID: (Human) P00973, (Rat) Q05961, (Mouse) P11928

          View more View less

          Entrez Gene ID: (Human) 4938, (Rat) 192281, (Mouse) 246730

          View more View less

          Function(s)
          2'-5'-oligoadenylate synthetase activity double-stranded RNA binding protein binding ATP binding nucleotidyltransferase activity
          Process(es)
          glucose metabolic process defense response response to virus positive regulation of interferon-beta production positive regulation of tumor necrosis factor production toll-like receptor 3 signaling pathway toll-like receptor 4 signaling pathway cellular response to interferon-alpha cellular response to interferon-beta glucose homeostasis defense response to bacterium surfactant homeostasis negative regulation of viral genome replication protein oligomerization defense response to virus type I interferon signaling pathway negative regulation of type I interferon-mediated signaling pathway regulation of ribonuclease activity interleukin-27-mediated signaling pathway cellular response to exogenous dsRNA positive regulation of monocyte chemotactic protein-1 production negative regulation of IP-10 production cellular response to virus antiviral innate immune response positive regulation of cellular respiration negative regulation of chemokine (C-X-C motif) ligand 2 production purine nucleotide biosynthetic process negative regulation of viral process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-lz9xs:80/100.66.72.150:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0