Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • OxPhos Blue Native WB Antibodies
          The product (cat # {{productObj.productId}}) that you're looking for has been discontinued and replaced with {{productObj.replacementProductId}}. The product (cat # {{productObj.productId}}) that you're looking for has been discontinued. Please contact Technical Support with any questions.

          Invitrogen

          OxPhos Blue Native WB Antibody Cocktail

          11 Published Figures
          10 References

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols
          Datasheet
          Certificates
          Protocols

          Cite OxPhos Blue Native WB Antibody Cocktail

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (11)
          OxPhos Blue Native WB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          OxPhos Blue Native WB Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          OxPhos Blue Native WB Antibody (45-7999) in ICC/IF

          Molecules and cells 2017 - Fig. 2 PINK1/Parkin-independent changes in ROS levels by NAM treatment Levels of mitochondria content (A) and superoxide (B) were determined using NAO or DHE at Day 1 or 2 of NAM treatment, and the effects were compared between normal fibroblasts and those whose Parkin expression was knocked down by siRNA (siParkin). The image of the protein blot (top) shows diminished Parkin protein levels in cells treated... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          OxPhos Blue Native WB Antibody in Immunocytochemistry (ICC/IF)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)
          OxPhos Blue Native WB Antibody in Western Blot (WB)

          Product Details

          45-7999

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          6 µg/mL
          View 9 publications 9 publications

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication
          Product Specifications

          Host/Isotype

          Mouse / Ig

          Class

          Cocktail

          Type

          Antibody

          Clone

          Cocktail

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.5 mg/mL

          Storage conditions

          4°C, do not freeze

          Shipping conditions

          Wet ice

          RRID

          AB_2533834

          Product Specific Information

          This Anti-OxPhos Complex Kit is a cocktail of monoclonal antibodies specific to human, bovine, mouse OxPhos Complex This antibody cocktail is validated for use in Western blotting.

          Mouse monoclonal [20C11B11B11] to (C-I) NDUFA9:

          Purification Method: IgG Fraction
          Amount: 120 µg
          Working Concentration: 2 µg/mL

          Mouse monoclonal [2E3GC12FB2AE2] to (C-II-70) SDHA:

          Purification Method: IgG Fraction
          Amount: 6 µg
          Working Concentration: 0.1 µg/mL

          Mouse monoclonal [13G12AF12BB11] to (C-III-Core 2) UQCRC2:

          Purification Method: IgG Fraction
          Amount: 60 µg
          Working Concentration: 1 µg/mL

          Mouse monoclonal [20E8C12] to (C-IV-subunit IV) COX IV:

          Purification Method: IgG Fraction
          Amount: 60 µg
          Working Concentration: 1 µg/mL

          Mouse monoclonal [15H4C4 ] to (C-V-alpha) ATP5A:

          Purification Method: Antigen Affinity Chromatography
          Amount: 60 µg
          Working Concentration: 1 µg/mL

          Target Information

          Altered levels of assembly can arise from mutations in individual subunits, mutations in assembly factors for the complex(es), mtDNA depletion or as a result of physiological and or pathological changes e.g. hormone treatment, exercise, diet or oxidative stress. Blue Native polyacrylamide gel electrophoresis (BN-PAGE) is a simple and effective way to subfractionate mitochondrial proteins as intact complexes on a single gel (in one dimension). It can be used to detect altered assembly of these complexes arising from mutations in subunits, mutations in assembly factors, or mtDNA depletion. This type of analysis has been performed with biopsy samples, platelets and fibroblast cells from patients with suspected mitochondrial diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0