    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Secondary Antibodies ›
          • Goat anti-Rabbit IgG Secondary Antibodies
          Invitrogen
          Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, PE
          2 Published Figures
          View all (247) Goat anti-Rabbit IgG antibodies
          Questions & Answers
          Cite Goat anti-Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody, PE
          Additional Information:
          {{banner.description}}
          • Published Figures (2)
          Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody (P-2771MP) in Flow
          Depletion of SK1 impaired cargo sorting in FMC-EV. A Depletion of SK1 expression in two SK1-knockout (SK1-KO) clones (C1 and C4) derived from FMC-1807 was verified by Western blotting analysis. B The concentration and size distribution of SK1-KO EV (right panel) were comparable with that of FMC-EV (left panel). Representative data of nanoparticle tracking analysis are shown. C Semi-quantitation of EV conten... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody in Flow Cytometry (Flow)
          Rabbit IgG (H+L) Cross-Adsorbed Secondary Antibody in Flow Cytometry (Flow)
          Product Details
          P-2771MP
          Applications
          Tested Dilution
          Immunocytochemistry (ICC/IF)
          Assay-Dependent
          Product Specifications
          Species Reactivity
          Rabbit
          Host/Isotype
          Goat / IgG
          Class
          Polyclonal
          Type
          Secondary Antibody
          Immunogen
          Gamma Immunoglobins Heavy and Light chains
          Conjugate
          PE PE PE
          • Alexa Fluor 350 
          • Alexa Fluor 405 
          • Alexa Fluor 430 
          • Alexa Fluor 488 
          • Alexa Fluor 514 
          • Alexa Fluor 532 
          • Alexa Fluor 546 
          • Alexa Fluor 555 
          • Alexa Fluor 568 
          • Alexa Fluor 594 
          • Alexa Fluor 633 
          • Alexa Fluor 635 
          • Alexa Fluor 647 
          • Alexa Fluor 660 
          • Alexa Fluor 680 
          • Alexa Fluor 700 
          • Alexa Fluor 750 
          • Alexa Fluor 790 
          • AP
          • APC 
          • Biotin 
          • Biotin-XX
          • Cyanine3
          • Cyanine5 
          • DyLight 350
          • DyLight 405
          • DyLight 488
          • DyLight 550
          • DyLight 594
          • DyLight 633
          • DyLight 650
          • DyLight 680
          • DyLight 755
          • DyLight 800
          • FITC
          • HRP
          • Pacific Blue
          • Pacific Orange
          • PE-Alexa Fluor 610
          • PE-Alexa Fluor 647
          • Rhodamine
          • Rhodamine Red-X
          • Texas Red
          • Texas Red-X
          • TRITC
          • Unconjugated
          • Request custom conjugation
          Excitation/Emission Max
          565/576 nm View spectra spectra
          Form
          Liquid
          Purification
          purified
          Storage buffer
          PBS, pH 7.5
          Contains
          5mM sodium azide
          Storage conditions
          4°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2539845
          Target
          IgG
          Cross Adsorption
          Against mouse IgG, mouse serum and human serum
          Antibody Form
          Whole Antibody
          Target Information
          Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
          View product citations for antibody P-2771MP on CiteAb
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-blue-7c59b9b899-7wltc:80/100.66.72.81:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline