    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • GFP Antibodies
          Invitrogen
          GFP Polyclonal Antibody
          9 Published Figures
          17 References
          View all (152) GFP antibodies
          Questions & Answers
          Cite GFP Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Published Figures (9)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          GFP Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 9
          GFP Antibody (PA1-9533) in ICC/IF
          Cells 2022 - Asp-GFP localization during kinetochore-driven microtubule regrowth (KDMTR) after colcemid-induced MT depolymerization. ( A ) S2 cells expressing both Asp-GFP and Cherry-tubulin were fixed and stained with anti-alpha-tubulin (red) and anti-GFP (green) antibodies and counterstained for DNA (DAPI, blue). Panel ( A1 ) shows an early KDMTR stage; panels ( A2 , A3 ) show more advanced KDMTR phases characterized ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunocytochemistry (ICC/IF)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Immunohistochemistry (IHC)
          GFP Antibody in Western Blot (WB)
          Product Details
          PA1-9533
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:2,000
          View 3 publications 3 publications
          Immunohistochemistry (IHC)
          -
          View 7 publications 7 publications
          Immunocytochemistry (ICC/IF)
          1:200-1:1,000
          View 7 publications 7 publications
          ELISA (ELISA)
          1:500-1:2,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Published species
          Tag
          Host/Isotype
          Chicken / IgY
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Purified recombinant Green Fluorescent fusion Protein.
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          500 µg
          Purification
          IgY fraction
          Storage buffer
          PBS
          Contains
          0.02% sodium azide
          Storage conditions
          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1074893
          Product Specific Information
          PA1-9533 detects Green Fluorescence Protein in N/A samples.
          PA1-9533 has been successfully used in Western blot, ELISA and immunocytochemistry procedures. By Western blot, PA1-9533 detects a 32 kDa protein representing Green Fluorescence Protein.
          PA1-9533 immunogen is a purified recombinant green fluorescence protein.
          Target Information
          The jellyfish Aequorea victoria contains green fluorescent protein (GFP Tag) that emits light in the bioluminescence reaction of the animal. GFP has been used widely as a reporter protein for gene expression in eukaryotic and prokaryotic organisms, and as a protein tag in cell culture and in multicellular organisms. As a fusion tag, GFP can be used to localize proteins, to study their movement or to research the dynamics of the subcellular compartments where these proteins are targeted. GFP technology has revealed considerable new insights in the physiological activities of living cells. GFP is a 27 kDa monomeric protein, which autocatalytically forms a fluorescent pigment. The wild type protein absorbs blue light (maximally at 395nm) and emits green light (peak emission 508nm) in the absence of additional proteins, substrates, or co-factors. GFP fluorescence is stable, species independent and is suitable for a variety of applications. GFP has been used extensively as a fluorescent tag to monitor gene expressin and protein localization. Moreover, other applications for GFP include its use in assessing protein protein interactions in the yeast two hybrid system, and in measuring distances between proteins in fluorescence energy transfer (FRET) experiments.
          Bioinformatics
          Protein Aliases: GFP; GFP tag; GFP2; green fluorescence; green fluorescent; green fluorescent protein; Turbo eGFP
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0