    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Glypican 5 Antibodies
          Invitrogen
          Glypican 5 Polyclonal Antibody
          View all (16) Glypican 5 antibodies
          Questions & Answers
          Cite Glypican 5 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Glypican 5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Glypican 5 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Glypican 5 Antibody (PA5-50185) in WB
          Western Blot analysis of Glypican 5 was performed using extracts from 293 cells. The membrane was probed with Glypican 5 Polyclonal Antibody (Product # PA5-50185). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Glypican 5 Antibody in Western Blot (WB)
          Glypican 5 Polyclonal Antibody
          Product Details
          PA5-50185
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:3,000
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Synthesized peptide derived from Internal of human GPC5.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 50% glycerol
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_2635638
          Product Specific Information
          The antibody detects endogenous levels of total Glypican 5 / GPC5 protein.
          Target Information
          Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation. [provided by RefSeq, Jul 2008].
          Bioinformatics
          Protein Aliases: bA93M14.1; glypican proteoglycan 5; Glypican-5; unnamed protein product
          View more View less
          Gene Aliases: A230034F01Rik; GPC5
          View more View less
          UniProt ID: (Human) P78333, (Mouse) Q8CAL5
          View more View less
          Entrez Gene ID: (Human) 2262, (Rat) 306157, (Mouse) 103978
          View more View less
          Function(s)
          protein binding molecular_function heparan sulfate proteoglycan binding
          Process(es)
          biological_process regulation of signal transduction cell migration positive regulation of canonical Wnt signaling pathway regulation of protein localization to membrane
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline