Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PACT Antibodies

          Proteintech

          PACT Polyclonal Antibody

          7 References
          View all (15) PACT antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PACT Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (12)
          PACT Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PACT Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PACT Antibody (10771-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol ) fixed HepG2 cells using 10771-1-AP (PACT antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PACT Antibody in Immunocytochemistry (ICC/IF)
          PACT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PACT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PACT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PACT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PACT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PACT Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          PACT Antibody in Western Blot (WB)
          PACT Antibody in Western Blot (WB)
          PACT Antibody in Western Blot (WB)
          PACT Antibody in Western Blot (WB)
          PACT Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          PACT Polyclonal Antibody

          Product Details

          10771-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 5 publications 5 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          0.5-4.0 µg
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PACT Fusion Protein Ag1214 (1-313 aa encoded by BC009470)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.17 mg/mL

          Amount

          25.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MSQSRHRAE APPLEREDSG TFSLGKMITA KPGKTPIQVL HEYGMKTKNI PVYECERSDV QIHVPTFTFR VTVGDITCTG EGTSKKLAKH RAAEAAINIL KANASICFAV PDPLMPDPSK QPKNQLNPIG SLQELAIHHG WRLPEYTLSQ EGGPAHKREY TTICRLESFM ETGKGASKKQ AKRNAAEKFL AKFSNISPEN HISLTNVVGH SLGCTWHSLR NSPGEKINLL KRSLLSIPNT DYIQLLSEIA KEQGFNITYL DIDELSANGQ YQCLAELSTS PITVCHGSGI SCGNAQSDAA HNALQYLKII AERK (1-313 aa encoded by BC009470)

          Target Information

          PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: dsRNA-binding protein; PKR associated protein X; PKR interacting protein; PKR-associated protein X; PKR-associating protein X; PRKRA; protein activator of the interferon-induced protein kinase; protein kinase, interferon-inducible double-stranded RNA-dependent activator; unnamed protein product

          View more View less

          Gene Aliases: DYT16; HSD14; lear; PACT; PRK; RAX

          View more View less

          UniProt ID: (Human) O75569, (Mouse) Q9WTX2, (Rat) Q4V8C7

          View more View less

          Entrez Gene ID: (Human) 8575, (Mouse) 23992, (Rat) 311130

          View more View less

          Function(s)
          RNA binding double-stranded RNA binding protein binding enzyme activator activity enzyme binding siRNA binding identical protein binding protein homodimerization activity pre-miRNA binding protein kinase binding protein kinase activator activity
          Process(es)
          immune response negative regulation of cell proliferation response to virus production of siRNA involved in RNA interference pre-miRNA processing production of miRNAs involved in gene silencing by miRNA protein stabilization regulation of production of small RNA involved in gene silencing by RNA small RNA loading onto RISC antiviral innate immune response protein phosphorylation positive regulation of cell proliferation cellular response to oxidative stress outer ear morphogenesis middle ear morphogenesis ear development skeletal system morphogenesis positive regulation of intrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-b6cf8ccb7-mmm2n:80/100.66.78.224:80.
          git-commit: 76b2fce788d0185f5e22c122d85c407206860d42
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.0-2026-offline-prod